Skip to Content
Merck
All Photos(1)

Key Documents

SML3395

Sigma-Aldrich

KDM5-C70

≥95% (HPLC)

Synonym(s):

C 70, C-70, C70, Ethyl 2-(((2-((2-(dimethylamino)ethyl)ethylamino)-2-oxoethyl)amino)methyl)-4-pyridinecarboxylate, Ethyl 2-(((2-((2-(dimethylamino)ethyl)ethylamino)-2-oxoethyl)amino)methyl)isonicotinate, KDM5-C49 ethyl ester, KDOAM-20 ethyl ester, KDOAM-21, KDOAM20 ethyl ester, KDOAM21

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H28N4O3
CAS Number:
Molecular Weight:
336.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

oil

color

, Light yellow to very dark red-bown

storage temp.

-10 to -25°C

SMILES string

CN(CCN(C(CNCC1=CC(C(OCC)=O)=CC=N1)=O)CC)C

InChI

1S/C17H28N4O3/c1-5-21(10-9-20(3)4)16(22)13-18-12-15-11-14(7-8-19-15)17(23)24-6-2/h7-8,11,18H,5-6,9-10,12-13H2,1-4H3

InChI key

WCILOMUUNVPIKQ-UHFFFAOYSA-N

Biochem/physiol Actions

KDM5-C70 (KDOAM-21) corresponds to the ethyl ester precursor of a selective αKG-competitive KDM5 histone demethylase inhibitor KDM5-C49 (KDOAM-21; KDM5A/B/C/D Ki = 2/1/6.1/3.4 nM vs. KDM4C/6B/3A/2A Ki = 0.51/4.55/2.59/4.4 μM; KDM5A/B/C/D IC50 = 1.1/0.8/3.2/2.7 μM by FDH assay with [αKG] = 1 mM & [E]/[S] = 0.5/15 μM; KDM5A/B/C IC50 = 25/30/59 nM by alphaLISA with [αKG] = 25 μM & [E]/[S] = 10/100 nM). KDM5-C70 treatment selectively upregulates cellular histone H3 trimethylation on Lys4 (H3K4me3), but not on Lys9/7/36 (5 μM, 3d), and inhibits KDM5-dependent cancer growth (by 85%/MCF7/11d, 97%/BT474/24d, and 70%/ZR-75-1/24d; 5 μM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural Basis for KDM5A Histone Lysine Demethylase Inhibition by Diverse Compounds
Cell Chemical Biology, 23(7), 769-781 (2016)
Lauren P Blair et al.
Science advances, 2(11), e1501662-e1501662 (2017-02-01)
The complexity by which cells regulate gene and protein expression is multifaceted and intricate. Regulation of 3' untranslated region (UTR) processing of mRNA has been shown to play a critical role in development and disease. However, the process by which
Stephanie B Hatch et al.
Epigenetics & chromatin, 10, 9-9 (2017-03-08)
Histone lysine demethylases (KDMs) are of interest as drug targets due to their regulatory roles in chromatin organization and their tight associations with diseases including cancer and mental disorders. The first KDM inhibitors for KDM1 have entered clinical trials, and
John R Horton et al.
Journal of medicinal chemistry, 61(7), 3193-3208 (2018-03-15)
Isomers of chiral drugs can exhibit marked differences in biological activities. We studied the binding and inhibitory activities of 12 compounds against KDM5A. Among them are two pairs of enantiomers representing two distinct inhibitor chemotypes, namely, ( R)- and (
Catrine Johansson et al.
Nature chemical biology, 12(7), 539-545 (2016-05-24)
Members of the KDM5 (also known as JARID1) family are 2-oxoglutarate- and Fe(2+)-dependent oxygenases that act as histone H3K4 demethylases, thereby regulating cell proliferation and stem cell self-renewal and differentiation. Here we report crystal structures of the catalytic core of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service