Skip to Content
Merck
All Photos(1)

Key Documents

SML3275

Sigma-Aldrich

Vamorolone

≥95% (HPLC)

Synonym(s):

(16α)-17,21-Dihydroxy-16-methylpregna-1,4,9(11)-triene-3,20-dione, (16α)-17,21-Dihydroxy-16-methylpregna-1,4,9(11)-triene-3,20-dione;, VBP 15, VBP15

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28O4
CAS Number:
Molecular Weight:
356.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.84

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O[C@]1([C@@]2([C@H]([C@@H]3CCC4=CC(=O)C=C[C@@]4(C3=CC2)C)C[C@H]1C)C)C(=O)CO

InChI

1S/C22H28O4/c1-13-10-18-16-5-4-14-11-15(24)6-8-20(14,2)17(16)7-9-21(18,3)22(13,26)19(25)12-23/h6-8,11,13,16,18,23,26H,4-5,9-10,12H2,1-3H3/t13-,16-,18+,20+,21+,22+/m1/s1

InChI key

ZYTXTXAMMDTYDQ-DGEXFFLYSA-N

Biochem/physiol Actions

Vamorolone, a synthetic steroid, is an orally available dissociative steroid with anti-inflammatory efficacy. It is a partial agonist of the glucocorticoid receptor (NR3C1) that appear to be safe and well tolerated. Also, vamorolone is a potent antagonist of the mineralocorticoid receptor (NR3C2). It is under development for children with Duchenne muscular dystrophy.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Liu et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(39), 24285-24293 (2020-09-13)
Duchenne muscular dystrophy is a genetic disorder that shows chronic and progressive damage to skeletal and cardiac muscle leading to premature death. Antiinflammatory corticosteroids targeting the glucocorticoid receptor (GR) are the current standard of care but drive adverse side effects
Christopher R Heier et al.
Life science alliance, 2(1) (2019-02-13)
Cardiomyopathy is a leading cause of death for Duchenne muscular dystrophy. Here, we find that the mineralocorticoid receptor (MR) and glucocorticoid receptor (GR) can share common ligands but play distinct roles in dystrophic heart and skeletal muscle pathophysiology. Comparisons of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service