Skip to Content
Merck
All Photos(1)

Key Documents

SML2769

Sigma-Aldrich

ASS234

≥98% (HPLC)

Synonym(s):

N,1-Dimethyl-5-[3-[1-(phenylmethyl)-4-piperidinyl]propoxy]-N-2-propyn-1-yl-1H-indole-2-methanamine, N-[[5-[3-(1-Benzylpiperidin-4-yl)propoxy]-1-methyl-1H-indol-2-yl]methyl]-N-methyl-2-propyn-1-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H37N3O
CAS Number:
Molecular Weight:
443.62
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN1C(CN(CC#C)C)=CC2=C1C=CC(OCCCC(CC3)CCN3CC4=CC=CC=C4)=C2

Biochem/physiol Actions

ASS234 is a brain-penetrant and orally active multi-target small molecule (MTSM) against (acetyl & butyryl) cholinesterases (hAChE/hBuChE IC50 = 0.81/1.82 μM), monoamine oxidases (hMAO-A/B IC50 = 0.27/120 nM), histamine H3 receptor (hH3R Ki = 84.2 nM; hH4R Ki >10 μM) and sigma-1/2 receptors (hS1R/rS2R = 2.82/50.3 nM). ASS234 exhibits antioxidative and neuroprotective effects in SH-SY5Y cultures (5 μM) and demonstrates therapeutic efficacy in neurodegerative disease models in vivo via sc. (0.62 mg/kg/d mice; 5 mg/kg rats), ip. (1 mg/kg mice) or po. (1 & 10 mg/kg mice).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Óscar M Bautista-Aguilera et al.
Journal of medicinal chemistry, 61(15), 6937-6943 (2018-07-04)
Contilisant, a permeable, antioxidant, and neuroprotectant agent, showing high nM affinity at H3R and excellent inhibition of the monoamine oxidases and cholinesterases, is an affine and selective S1R agonist in the nanomolar range, based on the binding affinity and functional
Upregulation of Antioxidant Enzymes by ASS234, a Multitarget Directed Propargylamine for Alzheimer's Disease Therapy.
Eva Ramos et al.
CNS neuroscience & therapeutics, 22(9), 799-802 (2016-07-07)
José Marco-Contelles et al.
Frontiers in neuroscience, 10, 294-294 (2016-07-23)
ASS2324 is a hybrid compound resulting from the juxtaposition of donepezil and the propargylamine PF9601N ASS2324 is a multi-target directed propargylamine able to bind to all the AChE/BuChE and MAO A/B enzymesASS2324 shows antioxidant, neuroprotective and suitable permeability propertiesASS2324 restores
Óscar M Bautista-Aguilera et al.
Angewandte Chemie (International ed. in English), 56(41), 12765-12769 (2017-09-02)
The therapy of complex neurodegenerative diseases requires the development of multitarget-directed drugs (MTDs). Novel indole derivatives with inhibitory activity towards acetyl/butyrylcholinesterases and monoamine oxidases A/B as well as the histamine H3 receptor (H3R) were obtained by optimization of the neuroprotectant
Javier Del Pino et al.
ACS chemical neuroscience, 9(12), 2880-2885 (2018-07-27)
There is clear evidence that neuroinflammation plays a crucial role in the pathogenesis of Alzheimer's disease. Consequently, modulating the inflammatory environment in brain has become a powerful and attractive strategy to deal with Alzheimer's disease physiopathology. In spite of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service