Skip to Content
Merck
All Photos(1)

Key Documents

SML2432

Sigma-Aldrich

Ethyl rosmarinate

≥98% (HPLC)

Synonym(s):

(αR)-α-[[(2E)-3-(3,4-Dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]-3,4-dihydroxy-benzenepropanoic acid ethyl ester, Rosmarinic acid ethyl ester, [R-(E)]-α-[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-3,4-dihydroxy-benzenepropanoic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20O8
CAS Number:
Molecular Weight:
388.37
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
protect from light

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(OCC)[C@H](OC(/C=C/C1=CC(O)=C(O)C=C1)=O)CC2=CC(O)=C(O)C=C2

InChI key

ROJRNQOAUDCMES-KRZKXXONSA-N

Biochem/physiol Actions

Ethyl rosmarinate is an anti-inflammatory and antioxidative compound present in Lycopus lucidus, Hyptis suaveolens and other medical plants that potently inhibits LPS induced production of nitric oxide and prostaglandin E2 in alveolar macrophages. Ethyl rosmarinate induces relaxation in aortic rings via an endothelium-independent pathway.
anti-inflammatory and antioxidative compound that potently inhibits LPS induced production of nitric oxide and prostaglandin E2 in alveolar macrophages

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handan Gokben Sevindik et al.
Chemical & pharmaceutical bulletin, 63(9), 720-725 (2015-09-04)
Thymus praecox ssp. grossheimii (RONNIGER) JALAS var. grossheimii (Lamiaceae) is used as an herbal tea for cold, stomachache, cough, and infections in Turkey. There are no phytochemical studies on this species. We performed phytochemical studies and quantitative analysis of rosmarinic
Piyawadee Wicha et al.
European journal of pharmacology, 766, 9-15 (2015-09-13)
Ethyl rosmarinate is an ester derivative of rosmarinic acid, a major constituent of Hyptis suaveolens. The present study investigated the vasorelaxant mechanism of ethyl rosmarinate in isolated rat aortic rings using an organ bath system. Ethyl rosmarinate (0.1 µM-3mM) produced
Hathairat Thammason et al.
European journal of pharmacology, 824, 17-23 (2018-02-03)
In this study, a series of rosmarinic acid and analogs were investigated for their anti-inflammatory potential against LPS-induced alveolar macrophages (MH-S). Our results showed that, among the test compounds, ethyl rosmarinate (3) exhibited the most potent inhibitory effect on NO

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service