Skip to Content
Merck
All Photos(1)

Key Documents

SML2358

Sigma-Aldrich

D-NMAPPD

≥98% (HPLC)

Synonym(s):

(1R, 2R)-N-myristoylamino-4′-nitro-phenylpropandiol-1, 3, (1R,2R)-B13, B13, N-[(1R,2R)-2-Hydroxy-1-(hydroxymethyl)-2-(4-nitrophenyl)ethyl]tetradecanamide, [R-(R*,R*)]-N-[2-hydroxy-1-(hydroxymethyl)-2-(4-nitrophenyl)ethyl]-tetradecanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H38N2O5
CAS Number:
Molecular Weight:
422.56
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -28 to -34, c = 0.1 in chloroform-d

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[N+](=O)([O-])c1ccc(cc1)[C@@H](O)[C@H](NC(=O)CCCCCCCCCCCCC)CO

InChI

1S/C23H38N2O5/c1-2-3-4-5-6-7-8-9-10-11-12-13-22(27)24-21(18-26)23(28)19-14-16-20(17-15-19)25(29)30/h14-17,21,23,26,28H,2-13,18H2,1H3,(H,24,27)/t21-,23-/m1/s1

InChI key

XUSDVLHKNBOGJY-FYYLOGMGSA-N

Biochem/physiol Actions

D-NMAPPD (B13), a myristoyl-CoA analog, is a cell penetrant inhibitor of N-myristoyltransferases that suppressed FRS2α (fibroblast growth factor receptor substrate 2) myristoylation. D-NMAPPD inhibits wild-type FGFRs and drug-resistant mutants (FGFRsDRM) oncogenic signaling in cancer cells. D-NMAPPD potently inhibits growth of xenograft tumors in mice. D-NMAPPD is a potent inhibitor of ceramidase that elevates endogenous ceramide levels and suppress growth of cancer cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mototeru Yamane et al.
Biochemistry and biophysics reports, 11, 174-181 (2017-09-29)
In this study, we showed that the dual addition of glucosyl ceramide synthase and ceramidase inhibitors to A549 cell culture led to the possibility of ceramide channel formation via endogenous palmitoyl-ceramide accumulation with an increase in cholesterol contents in the
Qianjin Li et al.
The Journal of biological chemistry, 293(17), 6434-6448 (2018-03-16)
Fibroblast growth factor (FGF)/FGF receptor (FGFR) signaling facilitates tumor initiation and progression. Although currently approved inhibitors of FGFR kinase have shown therapeutic benefit in clinical trials, overexpression or mutations of FGFRs eventually confer drug resistance and thereby abrogate the desired
Induction of Inflammation Disrupts the Negative Interplay between STING and S1P Axis That Is Observed during Physiological Conditions in the Lung.
Terlizzi, et al.
International Journal of Molecular Sciences, 24 (2023)
Michela Terlizzi et al.
Cells, 11(16) (2022-08-27)
Sphingosine-1-phosphate (S1P) is involved in inflammatory signaling/s associated with the development of respiratory disorders, including cancer. However, the underlying mechanism/s are still elusive. The aim of this study was to investigate the role of S1P on circulating blood cells obtained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service