Skip to Content
Merck
All Photos(1)

Key Documents

SML2145

Sigma-Aldrich

TD52

≥98% (HPLC)

Synonym(s):

N2,N3-bis(3-Ethynylphenyl)-2,3-quinoxalinediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16N4
CAS Number:
Molecular Weight:
360.41
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

C#CC1=CC=CC(NC2=NC3=CC=CC=C3N=C2NC4=CC=CC(C#C)=C4)=C1

InChI key

SCUPZFSEJFWQIS-UHFFFAOYSA-N

Biochem/physiol Actions

TD52, an erlotinib analog, is a putative inhibitor of CIP2A that exhibits potent antitumor efficacy on HCC and TNBC cells. TD52 induces apoptosis through downregulation of CIP2A (Cancerous inhibitor of protein phosphatase 2A) in HCC, NSCLC and TNBC cells. Apparently TD52 indirectly downregulates CIP2A transcripts through interrupting the binding of Elk1 to CIP2A promoter. TD52 is a weak inhibitor of EGFR tyrosine kinase.
putative inhibitor of CIP2A that exhibits potent antitumor efficacy on HCC and TNBC cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chun-Yu Liu et al.
European journal of cancer (Oxford, England : 1990), 72, 112-123 (2016-12-28)
Cancerous inhibitor of protein phosphatase 2A (CIP2A) has emerged as a therapeutic determinant mediating the anti-cancer effects of several new agents. We investigated the efficacy and mechanism of TD52, an erlotinib derivative with minimal p-EGFR inhibition but significant CIP2A downregulation
H-C Yu et al.
Cell death & disease, 5, e1359-e1359 (2014-08-01)
Protein phosphatase 2A (PP2A) is a tumor suppressor, which is functionally defective in various cancers. Previously, we found that PP2A activity determined the anticancer effect of bortezomib and erlotinib in hepatocellular carcinoma (HCC) cells. Here, we tested a novel erlotinib

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service