Skip to Content
Merck
All Photos(1)

Key Documents

SML1912

Sigma-Aldrich

GSK461364A

≥98% (HPLC)

Synonym(s):

(R)-5-(6-((4-methylpiperazin-1-yl)methyl)-1H-benzo[d]imidazol-1-yl)-3-(1-(2-(trifluoromethyl)phenyl)ethoxy)thiophene-2-carboxamide, 5-[6-[(4-Methyl-1-piperazinyl)methyl]-1H-benzimidazol-1-yl]-3-[(1R)-1-[2-(trifluoromethyl)phenyl]ethoxy]-2-thiophenecarboxamide, GSK461364

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H28F3N5O2S
CAS Number:
Molecular Weight:
543.60
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN1CCN(CC2=CC3=C(N=CN3C4=CC(O[C@@H](C5=C(C=CC=C5)C(F)(F)F)C)=C(C(N)=O)S4)C=C2)CC1

InChI

1S/C27H28F3N5O2S/c1-17(19-5-3-4-6-20(19)27(28,29)30)37-23-14-24(38-25(23)26(31)36)35-16-32-21-8-7-18(13-22(21)35)15-34-11-9-33(2)10-12-34/h3-8,13-14,16-17H,9-12,15H2,1-2H3,(H2,31,36)/t17-/m1/s1

InChI key

ZHJGWYRLJUCMRT-QGZVFWFLSA-N

Biochem/physiol Actions

GSK461364 is an imidazotriazine derivate that inhibits polo-like kinase 1 (Plk1) in a potent, reversible, ATP-competitive, and highly selective manner (Kiapp <0.5 nM vs. 0.86 μM for Plk2 and 200 μM for Plk3; ATP Kmapp = 16 μM), exhibiting much reduced or little potency toward a panel of 262 kinases. GSK461364A inhibits the growth of various cancer cultures (IG50 <100 nM) as a result of either growth arrest (cytostasis) or cytotoxicity in vitro and effectively suppresses the expansion of various human tumor exnografts in mice in vivo (25 or 50 mg/kg i.p. q2dx12).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aidan G Gilmartin et al.
Cancer research, 69(17), 6969-6977 (2009-08-20)
Polo-like kinase 1 (Plk1) is a conserved serine/threonine kinase that plays an essential role in regulating the many processes involved in mitotic entry and progression. In humans, Plk1 is expressed primarily during late G(2) and M phases and, in conjunction
Yongzhen Qian et al.
Clinical & experimental metastasis, 28(8), 899-908 (2011-09-29)
Few therapeutic strategies exist for the treatment of metastatic tumor cells in the brain because the blood-brain barrier (BBB) limits drug access. Thus the identification of molecular targets and accompanying BBB permeable drugs will significantly benefit brain metastasis patients. Polo-like
Yan Degenhardt et al.
Molecular cancer therapeutics, 9(7), 2079-2089 (2010-06-24)
Polo-like kinases are a family of serine threonine kinases that are critical regulators of cell cycle progression and DNA damage response. Predictive biomarkers for the Plk1-selective inhibitor GSK461364A were identified by comparing the genomics and genetics of a panel of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service