Skip to Content
Merck
All Photos(1)

Key Documents

SML1885

Sigma-Aldrich

Mitotane

≥98% (HPLC)

Synonym(s):

1-(2-Chlorophenyl)-1-(4-chlorophenyl)-2,2-dichloroethane, 1-Chloro-2-[2,2-dichloro-1-(4-chlorophenyl)ethyl]-benzene, 2,4′-DDD, o,p′-DDD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10Cl4
CAS Number:
Molecular Weight:
320.04
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=C(C(C(Cl)Cl)C2=CC=C(Cl)C=C2)C=CC=C1

InChI

1S/C14H10Cl4/c15-10-7-5-9(6-8-10)13(14(17)18)11-3-1-2-4-12(11)16/h1-8,13-14H

InChI key

JWBOIMRXGHLCPP-UHFFFAOYSA-N

Biochem/physiol Actions

Mitotane is a selective inhibitor of sterol-Oacyl-transferase 1 (SOAT1), also known as ACAT1, (acyl-coenzyme A cholesterol acyltransferase).
Mitotane is a selective inhibitor of sterol-Oacyl-transferase 1 (SOAT1), also known as ACAT1, (acyl-coenzyme A cholesterol acyltransferase). Mitotane inhibition of SOAT1 in adrenocortical carcinoma cells decreased the formation of cholesteryl esters, which increased free cholesterol levels and caused endoplasmic reticulum (ER) stress, the unfolded protein response, and eventually apoptosis.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthias Kroiss et al.
Hormones & cancer, 7(5-6), 345-355 (2016-11-01)
Mitotane is the only drug approved for treatment of the orphan disease adrenocortical carcinoma (ACC) and was recently shown to be the first clinically used drug acting through endoplasmic reticulum (ER)-stress induced by toxic lipids. Since mitotane has limited clinical
S Beloribi-Djefaflia et al.
Oncogenesis, 5, e189-e189 (2016-01-26)
Many human diseases, including metabolic, immune and central nervous system disorders, as well as cancer, are the consequence of an alteration in lipid metabolic enzymes and their pathways. This illustrates the fundamental role played by lipids in maintaining membrane homeostasis
Xiangyu Li et al.
iScience, 24(7), 102722-102722 (2021-07-15)
Clear cell renal cell carcinoma (ccRCC) is the most common histological type of kidney cancer and has high heterogeneity. Stratification of ccRCC is important since distinct subtypes differ in prognosis and treatment. Here, we applied a systems biology approach to
Silviu Sbiera et al.
Endocrinology, 156(11), 3895-3908 (2015-08-26)
Adrenocortical carcinoma (ACC) is a rare malignancy that harbors a dismal prognosis in advanced stages. Mitotane is approved as an orphan drug for treatment of ACC and counteracts tumor growth and steroid hormone production. Despite serious adverse effects, mitotane has
Christina Bothou et al.
Cancers, 13(16) (2021-08-28)
Current systemic treatment options for patients with adrenocortical carcinomas (ACCs) are far from being satisfactory. DNA damage/repair mechanisms, which involve, e.g., ataxia-telangiectasia-mutated (ATM) and ataxia-telangiectasia/Rad3-related (ATR) protein signaling or ribonucleotide reductase subunits M1/M2 (RRM1/RRM2)-encoded ribonucleotide reductase (RNR) activation, commonly contribute

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service