Skip to Content
Merck
All Photos(1)

Key Documents

SML1114

Sigma-Aldrich

IWP-4

≥95% (HPLC)

Synonym(s):

N-(6-Methyl-2-benzothiazolyl)-2-[[3,4,6,7-tetrahydro-3-(2-methoxyphenyl)-4-oxothieno[3,2-d]pyrimidin-2-yl]thio]-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H20N4O3S3
CAS Number:
Molecular Weight:
496.62
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 0.5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

O=C1C2=C(CCS2)N=C(SCC(NC3=NC(C=CC(C)=C4)=C4S3)=O)N1C5=C(OC)C=CC=C5

InChI

1S/C23H20N4O3S3/c1-13-7-8-14-18(11-13)33-22(24-14)26-19(28)12-32-23-25-15-9-10-31-20(15)21(29)27(23)16-5-3-4-6-17(16)30-2/h3-8,11H,9-10,12H2,1-2H3,(H,24,26,28)

InChI key

RHUJMHOIQBDFQR-UHFFFAOYSA-N

Biochem/physiol Actions

IWP-4 blocks Wnt family member 5a.[1] It blocks Wnt protein secretion and prevents the canonical and non-canonical signaling pathways.[2]
IWP-4 is a potent inhbitor of the Wnt pathway regulatory protein porupine (Porcn).
IWP-4 is a potent inhbitor of the Wnt pathway regulatory protein porupine (Porcn). IWP-4 blocks palmitylation, and subsquent secretion and activity of Wnt. IWP-4 induces the differentiation of human pluripotent stem cells into cardiomyocytes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microbioreactor Array Screening of Wnt Modulators and Microenvironmental Factors in Osteogenic Differentiation of Mesenchymal Progenitor Cells
Frith JE, et al.
PLoS ONE, 8(12), e82931-e82931 (2013)
Alla Narytnyk et al.
Stem cell reviews, 10(2), 316-326 (2014-01-09)
Here we provide a protocol for the directed differentiation of hEPI-NCSC into midbrain dopaminergic neurons, which degenerate in Parkinson's disease. hEPI-NCSC are neural crest-derived multipotent stem cells that persist into adulthood in the bulge of hair follicles. The experimental design
Qinghua Wang et al.
Communications biology, 5(1), 1018-1018 (2022-09-28)
The formation of wound epithelium initiates regeneration of amputated tail in Gekko japonicus. Energy metabolism is indispensable for the growth of living creatures and typically influenced by temperature. In this study, we reveal that low temperature lowers energy metabolism level
Oliver Schneider et al.
Materials today. Bio, 15, 100280-100280 (2022-05-24)
Over the last decade, Organ-on-Chip (OoC) emerged as a promising technology for advanced in vitro models, recapitulating key physiological cues. OoC approaches tailored for cardiac tissue engineering resulted in a variety of platforms, some of which integrate stimulation or probing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service