Skip to Content
Merck
All Photos(1)

Key Documents

SML1047

Sigma-Aldrich

GW2580

≥98% (HPLC)

Synonym(s):

5-[[3-Methoxy-4-[(4-methoxyphenyl)methoxy]phenyl]methyl]-2,4-Pyrimidinediamine, 5-[[3-Methoxy-4-[(4-methoxyphenyl)methoxy]phenyl]methyl]pyrimidine-2,4-diamine, GW 2580, GW-2580

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N4O3
CAS Number:
Molecular Weight:
366.41
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

Nc1nc(c(cn1)Cc2cc(c(cc2)OCc3ccc(cc3)OC)OC)N

InChI

1S/C20H22N4O3/c1-25-16-6-3-13(4-7-16)12-27-17-8-5-14(10-18(17)26-2)9-15-11-23-20(22)24-19(15)21/h3-8,10-11H,9,12H2,1-2H3,(H4,21,22,23,24)

InChI key

MYQAUKPBNJWPIE-UHFFFAOYSA-N

Application

GW2580 has been used to study its effect on paracrine signaling from stretched cardiomyocyte on cardiac fibroblast phenotype.

Biochem/physiol Actions

GW2580 is a cell-permeable, potent and selective ATP-competitive inhibitor of cFMS kinase, receptor for macrophage-colony stimulating factor, M-CSF or CSF-1, and a key regulator of macrophage and osteoclast activation and differentiation. FMS is a type III receptor tyrosine kinase that binds to the macrophage or monocyte colony-stimulating factor (M-CSF or CSF-1). Overexpression of CSF-1 and/or FMS has been implicated in a number of disease states including the growth of metastasis of some types of cancer, in promoting osteoclast proliferation in bone osteolysis, and in several inflammatory disorders. GW2580 selectively inhibits cFMS-mediated cellular functions in vitro at 0.06 μM as well as CSF-1-dependent tumor growth in vivo. GW2580 was inactive against 26 other kinases.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Rang Na et al.
Cell reports, 31(6), 107643-107643 (2020-05-14)
As current therapies benefit only a minority of cancer patients, additional therapeutic targets are needed. Tumor-associated macrophages (TAMs) have attracted attention for improving therapeutic responses, yet regulatory strategies remain elusive. Here, we show that the protein kinase A catalytic subunit
Kate M Herum et al.
Molecular biology of the cell, 28(14), 1871-1882 (2017-05-05)
Cardiac fibrosis is a serious condition currently lacking effective treatments. It occurs as a result of cardiac fibroblast (CFB) activation and differentiation into myofibroblasts, characterized by proliferation, extracellular matrix (ECM) production and stiffening, and contraction due to the expression of
Silvia Lonardi et al.
Cancer immunology research, 8(6), 829-841 (2020-04-03)
Langerhans cell histiocytosis (LCH) is a rare disorder characterized by tissue accumulation of CD1a+CD207+ LCH cells. In LCH, somatic mutations of the BRAF V600E gene have been detected in tissue LCH cells, bone marrow CD34+ hematopoietic stem cells, circulating CD14+
Desiree M Baron et al.
Human molecular genetics, 28(13), 2143-2160 (2019-02-27)
Aberrant translational repression is a feature of multiple neurodegenerative diseases. The association between disease-linked proteins and stress granules further implicates impaired stress responses in neurodegeneration. However, our knowledge of the proteins that evade translational repression is incomplete. It is also

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service