Skip to Content
Merck
All Photos(1)

Key Documents

SML0068

Sigma-Aldrich

CTP Inhibitor

≥98% (HPLC)

Synonym(s):

ZINC Compound 792949; 4-Chloro-3-[[(3-nitrophenyl)amino]sulfonyl]-benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9ClN2O6S
CAS Number:
Molecular Weight:
356.74
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥23 mg/mL

storage temp.

2-8°C

SMILES string

OC(=O)c1ccc(Cl)c(c1)S(=O)(=O)Nc2cccc(c2)[N+]([O-])=O

InChI

1S/C13H9ClN2O6S/c14-11-5-4-8(13(17)18)6-12(11)23(21,22)15-9-2-1-3-10(7-9)16(19)20/h1-7,15H,(H,17,18)

InChI key

IIJQJWNGBILZCU-UHFFFAOYSA-N

Application

CTP inhibitor may be used in cell signaling studies.
CTP inhibitor has been used in mouse to study the transition of endothelial to mesenchymal cells.

Biochem/physiol Actions

CTP inhibitor blocks the exchange of tricarboxylates the key intermediates in anabolism and catabolism by mitochondrial citrate transport protein (CTP).
CTP Inhibitor is an inhibitor of mitochondrial citrate transport protein, was the first purely competitive inhibitor to be discovered and is more potent than BTC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kishore S Malyavantham et al.
Chromosoma, 117(6), 553-567 (2008-07-05)
To study when and where active genes replicated in early S phase are transcribed, a series of pulse-chase experiments are performed to label replicating chromatin domains (RS) in early S phase and subsequently transcription sites (TS) after chase periods of
A metabolic basis for endothelial-to-mesenchymal transition
Xiong J, et al.
Molecular Cell, 69, 689-698 (2018)
Bowen Wu et al.
Cell metabolism, 32(6), 967-980 (2020-12-03)
Autoimmune T cells in rheumatoid arthritis (RA) have a defect in mitochondrial oxygen consumption and ATP production. Here, we identified suppression of the GDP-forming β subunit of succinate-CoA ligase (SUCLG2) as an underlying abnormality. SUCLG2-deficient T cells reverted the tricarboxylic acid (TCA)
Jiakang Sun et al.
Molecular and cellular pharmacology, 2(3), 101-110 (2010-08-06)
Cytoplasmic citrate is the prime carbon source for fatty acid, triacylglycerol, and cholesterol biosyntheses, and also regulates glucose metabolism via its allosteric inhibition of phosphofructokinase. It originates either via the efflux of citrate from the mitochondrial matrix on the inner
Marta Manco et al.
Biomolecules, 14(2) (2024-02-24)
Feline leukemia virus C receptor 1a (FLVCR1a), initially identified as a retroviral receptor and localized on the plasma membrane, has emerged as a crucial regulator of heme homeostasis. Functioning as a positive regulator of δ-aminolevulinic acid synthase 1 (ALAS1), the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service