Skip to Content
Merck
All Photos(1)

Key Documents

SBR00043

Sigma-Aldrich

PK150

Sorafenib analog

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H8ClF5N2O3
CAS Number:
Molecular Weight:
394.68
MDL number:
UNSPSC Code:
51112008
NACRES:
NA.76

Quality Level

form

powder

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

FC1(Oc2c(ccc(c2)NC(=O)Nc3cc(c(cc3)Cl)C(F)(F)F)O1)F

InChI

1S/C15H8ClF5N2O3/c16-10-3-1-7(5-9(10)14(17,18)19)22-13(24)23-8-2-4-11-12(6-8)26-15(20,21)25-11/h1-6H,(H2,22,23,24)

InChI key

SHPYXCLUNMOJOB-UHFFFAOYSA-N

General description

PK150 is a recently discovered small antimicrobial compound. It is a potent analog of sorafenib, an anticancer drug. PK150 has structural similarity with triclocarban (TCC) and triclosan (TCS) with a N,N′-diaryl urea motif in common.

Biochem/physiol Actions

PK150 is a potent antibiotic, which displays strong antibacterial activity against methicillin-resistant Staphylococcus aureus (MRSA) at a sub-micromolar level. It elicits anti-biofilm activity and eliminates persisters. PK150 also shows oral bioavailability, and in vivo efficacy in mouse model.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philipp Le et al.
Nature chemistry, 12(2), 145-158 (2019-12-18)
New drugs are desperately needed to combat methicillin-resistant Staphylococcus aureus (MRSA) infections. Here, we report screening commercial kinase inhibitors for antibacterial activity and found the anticancer drug sorafenib as major hit that effectively kills MRSA strains. Varying the key structural
Robert Macsics et al.
Applied and environmental microbiology, 86(16) (2020-06-07)
Triclocarban (TCC), a formerly used disinfectant, kills bacteria via an unknown mechanism of action. A structural hallmark is its N,N'-diaryl urea motif, which is also present in other antibiotics, including the recently reported small molecule PK150. We show here that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service