Skip to Content
Merck
All Photos(11)

Key Documents

SAB4503969

Sigma-Aldrich

Anti-phospho-PKA CAT (pThr197) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CAFD2, Anti-PKA C-beta, Anti-PKACB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 40 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr197)

Gene Information

Related Categories

Immunogen

The antiserum was produced against synthesized peptide derived from human PKA CAT around the phosphorylation site of Thr197.

Immunogen Range: 166-215

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seda Karabulut et al.
Cells, tissues, organs, 210(3), 218-238 (2021-07-29)
Ovarian hyperstimulation syndrome (OHSS) is one of the most common and iatrogenic complications of in vitro fertilization therapy, which is an exaggerated response to excess hormones resulting in the development of a large number of maturing follicles. Although the complications
Paula Ketilly Nascimento Alves et al.
Cells, 12(21) (2023-11-10)
Heart failure with preserved ejection fraction (HFpEF) is a complex syndrome associated with a high morbidity and mortality rate. Leucine supplementation has been demonstrated to attenuate cardiac dysfunction in animal models of cachexia and heart failure with reduced ejection fraction
Thirupura S Shankar et al.
Nature communications, 12(1), 4583-4583 (2021-07-30)
Voltage dependent anion channel 2 (VDAC2) is an outer mitochondrial membrane porin known to play a significant role in apoptosis and calcium signaling. Abnormalities in calcium homeostasis often leads to electrical and contractile dysfunction and can cause dilated cardiomyopathy and
Shanhong Fang et al.
Molecular medicine (Cambridge, Mass.), 29(1), 33-33 (2023-03-16)
Caveolin-1 has been predicted, based on RNA transcriptome sequencing, as a key gene in rotator cuff tear (RCT) and it is related to fatty infiltration. This study aims to elucidate the upstream and downstream mechanism of Caveolin-1 in fatty infiltration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service