Skip to Content
Merck
All Photos(8)

Key Documents

SAB4503879

Sigma-Aldrich

Anti-phospho-HSL (pSer552) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
IHC
WB
clone:
polyclonal
species reactivity:
mouse, rat, human
citations:
4
technique(s):
ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 116 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer552)

Gene Information

human ... LIPE(3991)

General description

Hormone sensitive lipase (HSL) is coded by LIPE (lipase E) gene. It is an intracellular lipase with substrates as triglycerides, diglycerides, monoglycerides, cholesterol esters and retinyl esters. HSL is mapped to human chromosome 19q13.

Immunogen

The antiserum was produced against synthesized peptide derived from human HSL around the phosphorylation site of Ser552.

Immunogen Range: 518-567

Application

Anti-HSL antibody has been used in western blotting.

Biochem/physiol Actions

Hormone sensitive lipase (HSL) plays an important role in lipolysis. Lack of HSL results in histologic abnormalities in white adipose tissue and an increase in macrophage infiltration.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Progesterone-induced down-regulation of hormone sensitive lipase (Lipe) and up-regulation of G0/G1 switch 2 (G0s2) genes expression in inguinal adipose tissue of female rats is reflected by diminished rate of lipolysis
Stelmanska E, et al.
The Journal of Steroid Biochemistry and Molecular Biology, 147, 31-39 (2015)
Identification of microdeletions spanning the Diamond-Blackfan anemia locus on 19q13 and evidence for genetic heterogeneity
Gustavsson P, et al.
American Journal of Human Genetics, 63(5), 1388-1395 (1998)
Null mutation in hormone-sensitive lipase gene and risk of type 2 diabetes
Albert JS, et al.
The New England Journal of Medicine, 370(24), 2307-2315 (2014)
Shanhong Fang et al.
Molecular medicine (Cambridge, Mass.), 29(1), 33-33 (2023-03-16)
Caveolin-1 has been predicted, based on RNA transcriptome sequencing, as a key gene in rotator cuff tear (RCT) and it is related to fatty infiltration. This study aims to elucidate the upstream and downstream mechanism of Caveolin-1 in fatty infiltration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service