Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500433

Sigma-Aldrich

Anti-Claudin 4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CLD4, CLDN4, CPE- receptor, Claudin-4, Clostridium perfringens enterotoxin receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
IHC
WB
clone:
polyclonal
species reactivity:
rat, mouse, human
citations:
5
technique(s):
ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 22 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLDN4(1364)

General description

The CLDN4 (claudin 4) gene is mapped to human chromosome 7q11.23. The encoded protein is an integral member, localized to tight junction. It is also expressed in the epithelial basolateral membrane.

Immunogen

The antiserum was produced against synthesized peptide derived from human Claudin 4.

Immunogen Range: 160-209

Biochem/physiol Actions

CLDN4 (claudin 4) is associated with epithelial differentiation, and is widely implicated in carcinomas. Its expression is considered to differentiate carcinoma with that of sarcoma. Claudin 4 expression is implicated in gastric cancer induced by Helicobacter pylori infection. It contributes to invasion, metastasis and prognosis of the disease. The CLDN4 gene is known to be overexpressed in a number of chemoresistant and metastatic cancer. Its localization to the tight junction prevents paracellular diffusion. Along with this, the extracellular loop interaction mediated by claudin 4 with the neighbouring cells makes it a differentiation marker.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of TGF-beta-induced Claudin-4 expression through c-Jun signaling in non-small cell lung cancer
Rachakonda G, et al.
Cellular Signalling, 28(10), 1537-1544 (2016)
Claudin-4 activity in ovarian tumor cell apoptosis resistance and migration
Hicks DA, et al.
BMC Cancer, 16(1), 788-788 (2016)
Claudin-4 expression distinguishes SWI/SNF complex-deficient undifferentiated carcinomas from sarcomas
Schaefer IM, et al.
Modern Pathology, 30(4), 539-539 (2017)
Claudin-1,-3 and-4 proteins and mRNA expression in benign and malignant breast lesions: a research study
Thokes AM, et al.
Breast Cancer Research, 7(2), R296-R296 (2005)
TLR2 activation induced by H. pylori LPS promotes the differential expression of claudin-4, -6, -7 and -9 via either STAT3 and ERK1/2 in AGS cells
Chavarria-Velazquez CO, et al.
Immunobiology, 223(1), 38-48 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service