Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200791

Sigma-Aldrich

Anti-Human IgG (Fab specific)-Peroxidase antibody, Mouse monoclonal

clone GG-6, purified from hybridoma cell culture

Synonym(s):

Anti-Human immunoglobulin G

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

peroxidase conjugate

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

GG-6, monoclonal

form

lyophilized powder

species reactivity

human

concentration

~2 mg/mL

technique(s)

direct ELISA: 1:16,000-1:32,000 using 2.5 μg/ml human IgG for coating.

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Human IgG (Fab specific)-Peroxidase antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the GG-6 hybridoma, produced by the fusion of mouse myeloma cells and splenocytes from mouse immunized with purified human IgG (Fab fragment). IgGs are the most common Immunoglobulins isotype in blood, lymph fluid, cerebrospinal fluid and peritoneal fluid and a key players in the humoral immune response. IgGs include four subclasses (IgG1, IgG2, IgG3, and IgG4), they consist of a variable Fab fragment (which includes the antibody recognition site), and a conserved Fc fragment. The IgG subclasses differ in their physical and chemical properties, their distribution pattern is found to be age-dependent, and every subclass has a specific biological function.

Immunogen

Purified human IgG (Fab Fragment)

Application

Direct ELISA: a working dilution of 1:16,000-1:32,000 is recommended using 2.5 μg/mL human IgG for coating.

Biochem/physiol Actions

Immunoglobulin G (IgG) participates in hypersensitivity type II and type III reactions. Maternal IgG is the only antibody transported across the placenta to the fetus. Maternal IgG passively immunizes the infants. IgG deficiencies are often associated with various diseases.

Physical form

Supplied as a lyophilized powder.

Other Notes

In order to obtain best results in different techniques and preparations we recommend determining optimal working concentration by titration test.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weizhong Li et al.
Nature communications, 14(1), 7114-7114 (2023-11-07)
SARS-CoV-2 is primarily transmitted through droplets and airborne aerosols, and in order to prevent infection and reduce viral spread vaccines should elicit protective immunity in the airways. The neonatal Fc receptor (FcRn) transfers IgG across epithelial barriers and can enhance
Human placental Fc receptors and the transmission of antibodies from mother to fetus.
Simister NE and Story CM
Journal of Reproductive Immunology, 37(1), 1-23 (1997)
Specific IgG for cat allergens in patients with allergic conjunctivitis.
Miyama A, et al.
International Ophthalmology, 35(4), 575-586 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service