Skip to Content
Merck
All Photos(3)

Key Documents

SAB2102759

Sigma-Aldrich

Anti-ZEB1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AREB6, Anti-BZP, Anti-MGC133261, Anti-NIL-2-A, Anti-Zinc finger E-box binding homeobox 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
WB
clone:
polyclonal
species reactivity:
dog, human, mouse, guinea pig, rabbit, rat
citations:
1
technique(s):
immunofluorescence: suitable
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

124 kDa

species reactivity

dog, human, mouse, guinea pig, rabbit, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunofluorescence: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZEB1(6935)

Immunogen

Synthetic peptide directed towards the N terminal region of human ZEB1

Biochem/physiol Actions

ZEB1 is a zinc finger transcription factor that represses T-lymphocyte-specific IL2 gene expression by binding to a negative regulatory domain 100 nucleotides 5-prime of the IL2 transcription start site.ZEB1 encodes a zinc finger transcription factor that represses T-lymphocyte-specific IL2 gene (MIM 147680) expression by binding to a negative regulatory domain 100 nucleotides 5-prime of the IL2 transcription start site (Williams et al., 1991 [PubMed 1840704]).[supplied by OMIM].

Sequence

Synthetic peptide located within the following region: KDDECESDAENEQNHDPNVEEFLQQQDTAVIFPEAPEEDQRQGTPEASGH

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ping Yin et al.
Cell communication and signaling : CCS, 18(1), 143-143 (2020-09-08)
Mesenchymal-like stemness is characterized by epithelial-mesenchymal transition (EMT). Breast cancer (BC) cell mesenchymal-like stemness is responsible for distal lung metastasis. Interrogation of databases showed that Fzd7 was closely associated with a panel of mesenchymal-related genes and a panel of stemness-related

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service