Skip to Content
Merck
All Photos(1)

Key Documents

SAB1401203

Sigma-Aldrich

Anti-IFNAR2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

IFN-R, IFN-alpha-REC, IFNABR, IFNARB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
human
citations:
2
technique(s):
western blot: 1 μg/mL

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IFNAR2(3455)

General description

The protein encoded by this gene is a type I membrane protein that forms one of the two chains of a receptor for interferons alpha and beta. Binding and activation of the receptor stimulates Janus protein kinases, which in turn phosphorylate several proteins, including STAT1 and STAT2. Multiple transcript variants encoding at least two different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

IFNAR2 (AAH02793.1, 1 a.a. ~ 331 a.a) full-length human protein.

Sequence
MLLSQNAFIVRSLNLVLMVYISLVFGISYDSPDYTDESCTFKISLRNFRSILSWELKNHSIVPTHYTLLYTIMSKPEDLKVVKNCANTTRSFCDLTDEWRSTHEAYVTVLEGFSGNTTLFSCSHNFWLAIDMSFEPPEFEIVGFTNHINVMVKFPSIVEEELQFDLSLVIEEQSEGIVKKHKPEIKGNMSGNFTYIIDKLIPNTNYCVSVYLEHSDEQAVIKSPLKCTLLPPGQESESAESAKIGGIITVFLIALVLTSTIVTLKWIGYICLRNSLPKVLRQGLTKGWNAVAIHRCSHNALQSETPELKQSSCLSFPSSWDYKRASLCPSD

Biochem/physiol Actions

The IFN (interferon) response is a part of the first line defense system against infection, which requires the binding of IFNs to the receptors (IFNAR1 and IFNAR2) for its activation. IFNAR2 (interferon αand βreceptor subunit 2) is associated with the JAK-STAT (Janus - signal transducer and activator of transcription) signaling pathway.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impaired expression of type I and type II interferon receptors in HCV-associated chronic liver disease and liver cirrhosis.
Chandra PK
PLoS ONE, 9(9), 1-10 (2014)
Type I Interferon Signaling Is Decoupled from Specific Receptor Orientation through Lenient Requirements of the Transmembrane Domain.
Sharma N
The Journal of Biological Chemistry, 291(7), 3371-3384 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service