Skip to Content
Merck
All Photos(4)

Key Documents

S7814

Solvent Safe pipette tips

volume range 1-200 μL, sterile

Synonym(s):

carbon fiber pipette tips, solvent safe pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
volume:
1—200 μL
sterility:
sterile
feature:
barrier
packaging:
pack of 384 ea (4 trays of 96 ea)
manufacturer/tradename:
Molecular BioProducts 5469
Pricing and availability is not currently available.

material

carbon fiber
colorless

sterility

sterile

feature

barrier

packaging

pack of 384 ea (4 trays of 96 ea)

manufacturer/tradename

Molecular BioProducts 5469

L

2 5/16 in. (58.8 mm)

maximum volume

200 μL

volume range

1-200 μL

volume

1—200 μL

compatibility

for use with

  • Biohit Variable 50-200 μL
  • Eppendorf Research/Reference 20-200 μL
  • Eppendorf Response 4850 20-500 μL
  • Eppendorf Research Pro 4860 20-300 μL
  • Eppendorf Titerman 4908 30-300 μL
  • Finnpipette 4027/4500
  • Digital Nichiryo BenchMate 20-200 μL
  • Pipetman P-200 20-200 μL
  • Rainin EDP-Series 250 (Limited Draw)
  • Transferpette

Looking for similar products? Visit Product Comparison Guide

General description

  • Solvent Safe carbon filter pipette tips
  • Filters block corrosive vapors created when pipetting volatile organic solvents such as TFA
  • Safely and accurately pipette phenol/chloroform DNA extractions
  • Perform peptide synthesis and HPLC analysis
  • Excellent alternative to glass pipettes

Other Notes

Browse all pipette tips

Legal Information

Solvent Safe is a trademark of Molecular BioProducts, Inc.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Could you please confirm if the Solvent Safe Pipette Tips, S7814, are suitable for use with xylene?

    1 answer
    1. The tips are made of polypropylene with a carbon fiber barrier. However, they may degrade or be damaged after repeated use with xylene.

      Helpful?

  2. Hey, Are the following solvent safe pipette tips product no.: S8064 + S7814 + S7939 compatible with Eppendorf research plus pipettes?

    1 answer
    1. All three tips state compatibility with Eppendorf Research Plus pipettors.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service