Skip to Content
Merck
All Photos(3)

Key Documents

S6197

Sigma-Aldrich

Anti-STIM1 (C-terminal) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-GOK, Anti-Stromal interaction molecule 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 90 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

immunoprecipitation (IP): 2.5-5 μg using extracts of rat PC12 cells.
indirect immunofluorescence: 2.5-5 μg/mL using human HeLa cells.
western blot: 2-4 μg/mL using whole extract of mouse 3T3 cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STIM1(6786)
mouse ... Stim1(20866)
rat ... Stim1(117086)

General description

Stromal interaction molecule 1 (STIM1) is a conserved single-pass transmembrane protein. STIM1 is expressed ubiquitously in a wide variety of human primary and transformed cell types. STIM1 localizes predominantly in the membrane of the endoplasmic reticulum (ER). It contains an N-terminal EF hand motif located in the ER lumen and appears to function as a sensor of ER Ca2+ levels.

Immunogen

synthetic peptide corresponding to amino acids 657-683 of human STIM1, conjugated to KLH via an N-terminal residue. The corresponding sequence is identical in rat, mouse, bovine, monkey, and dog.

Application

Anti-STIM1 (C-terminal) antibody produced in rabbit has been used in:
  • western blot
  • immunoprecipitation
  • immunofluorescence
  • immunohistochemistry

Biochem/physiol Actions

Stromal interaction molecule 1 (STIM1) is required for the activation of store-operated Ca2+ influx. STIM1 undergoes rapid oligomerization and redistributes into discrete spots (punctae) that move towards and accumulate in the cell periphery, possibly to activate Orai1 that is located in the plasma membrane. Overexpression of STIM1 and Orai1 together markedly increases the calcium release-activated channels (CRAC) current (I-CRAC). Dynamic assembly of TRPC1-STIM1-Orai1 ternary complex plays a major role in the activation of store-operated calcium (SOC) channel in response to internal Ca2+ store depletion.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

miR-424/322 regulates vascular smooth muscle cell phenotype and neointimal formation in the rat
Merlet E, et al.
Cardiovascular Research, 98(3), 458-468 (2013)
Elise Merlet et al.
Cardiovascular research, 98(3), 458-468 (2013-03-01)
Our aim was to identify new microRNAs (miRNAs) implicated in pathological vascular smooth muscle cells (VSMCs) proliferation and characterize their mechanism of action. MicroRNAs microarray and qRT-PCR results lead us to focus on miR-424 or its rat ortholog miR-322 (miR-424/322).
Zhen-Dong Zhu et al.
Cell death & disease, 9(2), 50-50 (2018-01-21)
Acute pancreatitis (AP) is an acute inflammatory process of the pancreas that is characterized by inflammation, edema, vacuolization and necrosis, which has significant morbidity and lethality. The pathogenesis of AP has not been established completely. An early and critical feature
STIM1 is a microtubule plus end tracking protein involved in remodeling of the endoplasmic reticulum
Grigoriev I, et al.
Current Biology, 18(3), 177-177 (2008)
SOCE induced calcium overload regulates autophagy in acute pancreatitis via calcineurin activation
Zhu ZD, et al.
Cell Death & Disease, 9(2), 50-50 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service