Skip to Content
Merck
All Photos(1)

Key Documents

PZ0152

Sigma-Aldrich

UK-356618

≥98% (HPLC)

Synonym(s):

(2R)-N1-[(1S)-2,2-Dimethyl-1-({[(1R)-1-phenylethyl]amino}carbonyl)propyl]-2-{3-[(3-methyl-4-phenyl)-phenyl]propyl}-(N4-hydroxy)butanediamide, UK-356,618

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H43N3O4
CAS Number:
Molecular Weight:
557.72
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D >+25.0°, c = 0.5 in methanol

color

white to tan

solubility

DMSO: ≥25 mg/mL

storage temp.

2-8°C

SMILES string

C[C@@H](NC(=O)[C@@H](NC(=O)[C@H](CCCc1ccc(c(C)c1)-c2ccccc2)CC(=O)NO)C(C)(C)C)c3ccccc3

InChI

1S/C34H43N3O4/c1-23-21-25(19-20-29(23)27-16-10-7-11-17-27)13-12-18-28(22-30(38)37-41)32(39)36-31(34(3,4)5)33(40)35-24(2)26-14-8-6-9-15-26/h6-11,14-17,19-21,24,28,31,41H,12-13,18,22H2,1-5H3,(H,35,40)(H,36,39)(H,37,38)/t24-,28-,31-/m1/s1

InChI key

JJHRUUKMPWUYIB-HVOSOHGQSA-N

Application

UK-356618 has been used as a specific matrix metalloproteinase 3 (MMP3) inhibitor to study its effect on amitriptyline-induced glial cell line-derived neurotrophic factor (GDNF) mRNA expression and to confirm its influence on amitriptyline-induced zymographic MMP-9 activation in rat C6 astroglial cells.

Biochem/physiol Actions

UK 356618 has the potential to inhibit interleukin (IL)-6-induced cell migration in NCI H446 small cell lung cancer cells in vitro.
UK-356,618 is a potent, selective inhibitor of Matrix metalloproteinase MMP3 (stromelysin-1). MMP-3 IC50 = 5.9 nM, selectivity is 140-fold vs MMP-1, -2, -9, and -14.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Na Jiang et al.
Oncotarget, 6(38), 40719-40733 (2015-11-04)
Our previous studies show that the phosphorylation of ataxia-telangiectasia mutated (ATM) induced by interleukin 6 (IL-6) treatment contributes to multidrug resistance formation in lung cancer cells, but the exact role of ATM activation in IL-6 increased metastasis is still elusive.
Hiromi Abe et al.
Biochemical and biophysical research communications, 479(4), 907-912 (2016-09-30)
Neurotrophic/growth factors derived from glial cells, especially astrocytes, have been implicated in mood disorders and the pharmacological effects of antidepressant drugs. Previous studies demonstrated that the release of glial cell line-derived neurotrophic factor (GDNF) induced by the tricyclic antidepressant amitriptyline

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service