Skip to Content
Merck
All Photos(2)

Key Documents

PLA0123

Sigma-Aldrich

Rabbit anti-ErbB2 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

CD antigen CD340, CD340, HER-2, HER-2/neu, HER2, MLN 19, NEU, NGL, TKR1, c-erb B2/neu protein, herstatin, human epidermal growth factor receptor 2, metastatic lymph node gene 19 protein, neuro/glioblastoma derived oncogene homolog, neuroblastoma/glioblastoma derived oncogene homolog, p185erbB2, proto-oncogene Neu, proto-oncogene c-ErbB-2, tyrosine kinase-type cell surface receptor HER2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunohistochemistry: 1:100- 1:500
immunoprecipitation (IP): 1-4 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_004439.2

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... ErbB2(2064)

Immunogen

The epitope recognized by PLA0123 maps to a region between residue 1200 and the C-terminus (residue 1255) of human v-erb-b2 erythroblastic leukemia viral oncogene homolog 2 using the numbering given in entry NP_004439.2 (GeneID 2064).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

ErbB2 is a member of the receptor tyrosine kinase family of proteins. The four members of this family are ErbB1 (Epidermal growth factor receptor; EGFR/HER1), ErbB2 (HER2/Neu), ErbB3 (HER3), and ErbB4 (HER4). The ErbB receptor members are single-pass transmembrane proteins with an extracellular ligand-binding domain and a cytoplasmic tyrosine-kinase domain. The epidermal growth factor (EGF) and its related proteins serve as the ligands for the ErbB receptors. Upon binding, the receptors heterodimerize, autophosphorylate and make contact with adapter molecules to transmit cellular signals involved in cell proliferation, survival, and differentiation. The ErbB2 receptor is the only member that does not interact with a ligand, but instead functions as a heterodimeric partner to coordinate signals from other ligand bound ErbB receptors. Amplification and/or overexpression of ErbB2 have been reported in many cancers.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service