Skip to Content
Merck
All Photos(2)

Documents

P9122

Sigma-Aldrich

Anti-Purinergic Receptor P2X7 (extracellular) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-P2Z

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human, rat, mouse

technique(s)

western blot: 1:200 using rat brain lysate and human cell lines

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... P2RX7(5027)
mouse ... P2rx7(18439)
rat ... P2rx7(29665)

General description

P2X receptors are membrane-bound ion channels that are activated by extracellular ATP. These receptors consist of seven known isoforms ranging from P2X1 to P2X7 . P2X7 modulates several cellular functions such as cell death, cytokine release and mRNA synthesis . Anti-Purinergic Receptor P2(X7) (extracellular) antibody is specific for P2X7 in rats and mice. The antibody also reacts with human K562 (Chronic myelogeneous leukemia); WEHI-231 (Lymphoma, B lymphocyte) and HL-60 (promyelocytic leukemia) cell lines.

Immunogen

Synthetic peptide corresponding to residues 136-152 of mouse P2X7 receptor. This antigenic epitope is identical in humans and rats; whereas in bovines, only 14/17 residues are identical.

Application

Anti-Purinergic Receptor P2(X7) (extracellular) antibody is suitable for use in immunoblot and western blot (1:200, using rat brain lysate and human cell lines).

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, 1% BSA, and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular Physiology of P2X Receptors.
North, R., A.
Physical Review, 82(4), 1013-1067 (2002)
Y Ding et al.
Journal of the autonomic nervous system, 81(1-3), 289-294 (2000-06-28)
A role for ATP in nociception and pain induction was proposed on the basis of human psychophysical experiments shortly after the formulation of the purinergic hypothesis. Following the pharmacological definition of distinct P2X and P2Y purinergic receptor subtypes by Burnstock
Audrey Cambon et al.
Frontiers in immunology, 14, 1228122-1228122 (2023-12-11)
IL-1β is a leaderless cytokine with poorly known secretory mechanisms that is barely detectable in serum of patients, including those with an IL-1β-mediated disease such as systemic juvenile idiopathic arthritis (sJIA). Leukocyte microvesicles (MVs) may be a mechanism of IL-1β
Letizia Penolazzi et al.
Frontiers in cell and developmental biology, 11, 1180774-1180774 (2023-05-22)
Identifying the subcellular localization of a protein within a cell is often an essential step in understanding its function. The main objective of this report was to determine the presence of the P2X7 receptor (P2X7R) in healthy human cells of
Alba Clara Sarti et al.
Function (Oxford, England), 2(2), zqab005-zqab005 (2021-01-28)
Basal expression of the P2X7 receptor (P2X7R) improves mitochondrial metabolism, Adenosine 5'-triphosphate (ATP) synthesis, and overall fitness of immune and non-immune cells. We investigated P2X7R contribution to energy metabolism and subcellular localization in fibroblasts (mouse embryo fibroblasts and HEK293 human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service