Skip to Content
Merck
All Photos(1)

Key Documents

P6557

Sigma-Aldrich

Carestream® Processing chemicals

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
45141500
NACRES:
NB.35
Pricing and availability is not currently available.

form

solid

Looking for similar products? Visit Product Comparison Guide

Reconstitution

Packet of powder to make the indicated volume.

Legal Information

Kodak is a registered trademark of Eastman Kodak Company

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katlijn Vints et al.
Scientific reports, 9(1), 130-130 (2019-01-16)
Analysis of neuronal arborization and connections is a powerful tool in fundamental and clinical neuroscience. Changes in neuronal morphology are central to brain development and plasticity and are associated with numerous diseases. Golgi staining is a classical technique based on
Provas Das et al.
Scientific reports, 5, 10395-10395 (2015-05-21)
Enveloped viruses enter host cells through membrane fusion and the cells in turn alter their shape to accommodate components of the virus. However, the role of nonmuscle myosin II of the actomyosin complex of host cells in membrane fusion is

Questions

  1. Is this product for preparing fixer and developer or what products does it include?

    1 answer
    1. Product P6557 contains only one component: the autoradiography solution fixer, in powder form, which can be made up to 1 gallon (3.785 liters) of solution by dissolving it. It doesn't contain the developer.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service