Skip to Content
Merck
All Photos(1)

Key Documents

P6024

Sigma-Aldrich

Putrescine dihydrochloride

γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture

Synonym(s):

1,4-Butanediamine dihydrochloride, 1,4-Diaminobutane dihydrochloride, Tetramethylenediamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)4NH2 · 2HCl
CAS Number:
Molecular Weight:
161.07
Beilstein:
3906680
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

sterility

γ-irradiated

product line

BioXtra

Assay

≥97% (HPLC)

form

lyophilized powder

technique(s)

cell culture | mammalian: suitable

mp

280 °C (dec.) (lit.)

solubility

cell culture medium: 0.16 mg/mL

storage temp.

room temp

SMILES string

Cl[H].Cl[H].NCCCCN

InChI

1S/C4H12N2.2ClH/c5-3-1-2-4-6;;/h1-6H2;2*1H

InChI key

XXWCODXIQWIHQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Binds to the polyamine modulatory site of the NMDA receptor and potentiates NMDA-induced currents; precursor of spermidine.
Binds to the polyamine modulatory site of the NMDA receptor; potentiates NMDA-induced currents.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Tao et al.
Human reproduction (Oxford, England), 30(8), 1867-1875 (2015-06-18)
Does peri-ovulatory putrescine supplementation of older mice improve oocyte quality and reduce the incidence of embryo resorption? Peri-ovulatory putrescine supplementation in older mice improved oocyte quality, as indicated by increased blastocyst cell numbers and reduced the incidence of embryo resorption.
Injae Hwang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(6), 2397-2411 (2015-02-26)
Firmicutes and Bacteroidetes, 2 major phyla of gut microbiota, are involved in lipid and bile acid metabolism to maintain systemic energy homeostasis in host. Recently, accumulating evidence has suggested that dietary changes promptly induce the alteration of abundance of both
Quan Findlay et al.
American journal of physiology. Gastrointestinal and liver physiology, 307(7), G741-G748 (2014-08-16)
Gut motility disorders can result from an absent, damaged, or dysfunctional enteric nervous system (ENS). Cell therapy is an exciting prospect to treat these enteric neuropathies and restore gut motility. Previous studies have examined a variety of sources of stem/progenitor
Takahiro Takayama et al.
Analytica chimica acta, 898, 73-84 (2015-11-04)
Chiral metabolites are found in a wide variety of living organisms and some of them are understood to be physiologically active compounds and biomarkers. However, the overall analysis of chiral metabolomics is quite difficult due to the high number of
Tadeu Chaves de Figueiredo et al.
Talanta, 142, 240-245 (2015-05-25)
A quantitative and confirmatory high-performance liquid chromatography with ultraviolet detection (HPLC-UV) method for the determination of bioactive amines in the albumen and yolk of commercial eggs was developed, optimized and validated by analyte extraction with trichloroacetic acid and pre-column derivatization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service