Skip to Content
Merck
All Photos(1)

Key Documents

P2906

Sigma-Aldrich

Poly(ethylene glycol)

Hybri-Max, mol wt 3,000-3,700, waxy solid, BioReagent, suitable for hybridoma

Synonym(s):

Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12161902
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic (organic)

Quality Level

grade

Hybri-Max

sterility

autoclaved

product line

BioReagent

form

waxy solid

mol wt

3,000-3,700

technique(s)

cell culture | hybridoma: suitable

impurities

endotoxin, tested

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) is a non-ionic hydrophilic polymer and is available in different molecular weights. It either exists in the form of a linear or branched structure. It aids in the purification and crystal growth of proteins and nucleic acids. PEG along with dextran is used to achieve an aqueous polymer two-phase system, which is essential for the purification of biological materials. It permits cell fusion through its interaction with the cell membrane.

Application

Poly(ethylene glycol) has been used in the production of monoclonal antibodies.
Recommended for cell fusion.

Legal Information

Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harris JM
Poly(Ethylene Glycol) Chemistry: Biotechnical and Biomedical Applications (2013)
Extensive nuclear localization of alpha-synuclein in normal rat brain neurons revealed by a novel monoclonal antibody
Yu S, et al.
Neuroscience journal, 145(2), 539-555 (2007)
Agathe Dubuisson et al.
Cell death & disease, 10(2), 101-101 (2019-02-06)
Development of therapeutic antibodies in oncology has attracted much interest in the past decades. More than 30 of them have been approved and are being used to treat patients suffering from cancer. Despite encouraging results, and albeit most clinical trials
Mark A Rice et al.
Acta biomaterialia, 5(1), 152-161 (2008-09-17)
Ultrasound has potential as a non-destructive analytical technique to provide real-time online assessments of matrix evolution in cell-hydrogel constructs used in tissue engineering. In these studies, chondrocytes were encapsulated in poly(ethylene glycol) hydrogels, and gel degradation was manipulated to provide
Idalis Villanueva et al.
Acta biomaterialia, 5(8), 2832-2846 (2009-06-11)
The pericellular matrix (PCM) surrounding chondrocytes is thought to play an important role in transmitting biochemical and biomechanical signals to the cells, which regulates many cellular functions including tissue homeostasis. To better understand chondrocytes interactions with their PCM, three-dimensional poly(ethylene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service