Skip to Content
Merck
All Photos(1)

Key Documents

N8638

Sigma-Aldrich

4-Nitrophenyl β-D-N,N′,N′′-triacetylchitotriose

chitinase substrate; lysozyme substrate, ≥98% (TLC), powder

Synonym(s):

4-Nitrophenyl b-D-N,N′,N′′-triacetylchitotriose, 4-Nitrophenyl beta-D-N,N′,N′′-triacetylchitotriose, N-[2-[5-acetamido-6-[5-acetamido-4-hydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy-4-hydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H44N4O18
CAS Number:
Molecular Weight:
748.69
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Product Name

4-Nitrophenyl β-D-N,N′,N′′-triacetylchitotriose, chitinase and lysozyme substrate

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

DMSO: 9.80-10.20 mg/mL, clear to slightly hazy, colorless to yellow

storage temp.

2-8°C

SMILES string

CC(=O)NC1C(O)C(O)C(CO)OC1OC2C(O)C(NC(C)=O)C(OC2CO)OC3C(O)C(NC(C)=O)C(OC3CO)Oc4ccc(cc4)N(=O)=O

InChI

1S/C30H44N4O18/c1-11(38)31-19-23(42)22(41)16(8-35)48-29(19)51-27-18(10-37)50-30(21(25(27)44)33-13(3)40)52-26-17(9-36)49-28(20(24(26)43)32-12(2)39)47-15-6-4-14(5-7-15)34(45)46/h4-7,16-30,35-37,41-44H,8-10H2,1-3H3,(H,31,38)(H,32,39)(H,33,40)

InChI key

UWYYAJFSFHIFNY-UHFFFAOYSA-N

General description

4-Nitrophenyl β-D-N,N′,N′′-triacetylchitotriose is a substrate for hydrolytic activity of chitinase enzyme, with the release of chromogenic end product p-nitrophenol, measured at 405 nm.[1][2]

Application

4-Nitrophenyl β-D-N,N′,N′′-triacetylchitotriose has been used as a substrate for determining the endochitinase activity of a novel protein isolated from the yam (Dioscorea opposita Thunb.) (DOI),[1] enzyme activity of recombinant chitinase CvChi45.[2]

Substrates

Chitinase and lysozyme substrate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression and efficient secretion of a functional chitinase from Chromobacterium violaceum in Escherichia coli
Lobo MDP, et al.
BMC biotechnology, 13(1), 46-46 (2013)
Osawa, T. and Nakazawa, Y.
Biochimica et Biophysica Acta, 130, 56-56 (1966)
Kam Lok Wong et al.
Scientific reports, 5, 10179-10179 (2015-07-15)
A novel protein, designated as DOI, isolated from the Chinese yam (Dioscorea opposita Thunb.) could be the first protein drug for the treatment of menopausal syndrome and an alternative to hormone replacement therapy (HRT), which is known to have undesirable

Questions

  1. What is the solubility of this substrate in water? (4-Nitrophenyl β-D-N,N′,N′′-triacetylchitotriose)

    1 answer
    1. The solubility of N8638 (4-Nitrophenyl β-D-N,N′,N′′-triacetylchitotriose) in water was not tested in-house. Its solubility in DMSO is 9.80-10.20 mg/mL.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service