Skip to Content
Merck
All Photos(2)

Key Documents

M0445

Sigma-Aldrich

Anti-MDMX antibody, Mouse monoclonal

enhanced validation

clone MDMX-82, purified from hybridoma cell culture

Synonym(s):

Anti-HDMX, Anti-MDM4, Anti-p53 Binding Protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MDMX-82, monoclonal

form

buffered aqueous solution

mol wt

antigen ~80 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
microarray: suitable
western blot: 1-2 μg/mL using total cell extract of HEK 293T cells transfected with human MDMX

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MDM4(4194)
mouse ... Mdm4(17248)
rat ... Mdm4(304798)

General description

Double minute 4 protein or MDMX is a nuclear protein. The gene encoding it is localized on human chromosome 1q32.1.
Monoclonal Anti-MDMDX (mouse IgG1 isotype) is derived from the hybridoma MDMX-82 produced by the fusion of mouse myeloma cells (NS1 cells) and splenocytes from BALB/c mice. MDMX belongs to the MDM family. The MDMX/ mouse double minute 4 (MDM4) gene encode a 490 amino acid protein. MDMX comprises a putative nuclear localization signal, a really interesting new gene (RING) finger domain in the C terminal and a p53-binding domain in the N terminal. MDMX has significant structural similarity to MDM2 E3 ubiquitin ligase.

Specificity

Monoclonal Anti-MDMX recognizes human MDMX.

Application

Monoclonal Anti-MDMX antibody is suitable for use in immunocytochemistry, immunoprecipitation, immunoblot (approx. 80 kDa), microarray, and ELISA. The product may also be used for western blot and for assessing ectopic expression of MDMX (using a conc. of 1-2 μg/mL in total cell extract of HEK 293T cells transfected with human MDMX).

Biochem/physiol Actions

MDMX is a structural homolog of MDM2 and inhibits the tumor suppressor, p53. MDMX also prevents MDM2-mediated degradation of p53, and subsequently regulates cellular levels of p53. MDMX also modulates cell growth and survival. Thus, MDMX can be useful as a therapeutic target in cancers, especially in tumors where native p53 functions can be restored . Monoclonal Anti-MDMX antibody is specific for human MDMX.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinna Zhang et al.
Cancer research, 69(20), 7960-7968 (2009-10-08)
MdmX and Mdm2 regulate p53 tumor suppressor functions by controlling p53 transcriptional activity and/or stability in cells exposed to DNA damage. Accumulating evidence indicates that ATM-mediated phosphorylation and degradation of Mdm2 and MdmX may be the initial driving force that
M W Jackson et al.
Molecular and cellular biology, 20(3), 1001-1007 (2000-01-11)
The p53 tumor suppressor protein is stabilized in response to cellular stress, resulting in activation of genes responsible for either cell cycle arrest or apoptosis. The cellular pathway for releasing normal cells from p53-dependent cell cycle arrest involves the Mdm2
Emilie A Chapeau et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(12), 3151-3156 (2017-03-08)
Inhibitors of double minute 2 protein (MDM2)-tumor protein 53 (TP53) interaction are predicted to be effective in tumors in which the TP53 gene is wild type, by preventing TP53 protein degradation. One such setting is represented by the frequent CDKN2A
In vitro and in silico studies of MDM2/MDMX isoforms predict Nutlin-3A sensitivity in well/de-differentiated liposarcomas
Bozzi F, et al.
Laboratory Investigation; a Journal of Technical Methods and Pathology, 93(11), 1-9 (2013)
F Mancini et al.
Current genomics, 10(1), 42-50 (2009-09-02)
MDM family proteins are crucial regulators of the oncosuppressor p53. Alterations of their gene status, mainly amplification events, have been frequently observed in human tumors.MDM4 is one of the two members of the MDM family. The human gene is located

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service