I3018
IEF mix 3.6-9.3
lyophilized powder
Synonym(s):
isoelectric focusing marker, protein markers
About This Item
Recommended Products
Application
Other Notes
Reconstitution
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service