Key Documents
HS9913
Cardboard Slide Tray
Holds 20 x slides, blue, trim
About This Item
UNSPSC Code:
41123302
NACRES:
NB.35
Pricing and availability is not currently available.
material
cardboard
feature
Holds 20 x slides
manufacturer/tradename
Heathrow Scientific HS9913
size
208x341x9 mm
color
blue, trim
Features and Benefits
  • Easily organize slides with the index on the front cover that corresponds with the number grid inside
  • Recessed slide compartments and inside covers protect specimens. Slides cannot be touched when the cover is closed, reducing contamination
  • Front protection flaps are attached with durable tape for repeated use. Fold flaps flat under the slide holder to save space when in use
  • Thumb cut in slide compartments enables easy removal of slides with slight pressure at one end
  • Compartment holder is made from a double layer of densely pressed cardboard. Cover is a single layer cardboard
  • Disposable
Choose from one of the most recent versions:
Certificates of Analysis (COA)