Key Documents
HS20632A
Pipette Filler
black
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Pricing and availability is not currently available.
material
black
black natural rubber
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS20632A
L × W × H
13 cm × 5.4 cm × 5.4 cm
capacity
~35 mL
General description
The natural rubber Pipette Filler is great for use in one-handed operation.
Features and Benefits
  • Efficient one-handed operation
  • Control air-evacuation, liquid uptake, and liquid dispensing with three valves each containing chemical-resistant stainless steel balls in valves
  • Pinch release-valve for smooth liquid flow
  • 5.1 in L x 2.1 in W x 2.1 in H (13 cm L x 5.4 cm W x 5.4 cm H)
Choose from one of the most recent versions:
Certificates of Analysis (COA)