HS120052
DuraPorter® specimen transport box
clear, with blue handles
Synonym(s):
multichannel pipette reservoir, reagent reservoir
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41123300
NACRES:
NB.17
Recommended Products
material
polycarbonate
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120052
size
380x196x160 mm
color
clear, with blue handles
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The DuraPorter Specimen or Sample Transport Box is ideal for transporting biological and clinical samples, as well as instruments and products that you want to keep clean and dry under testing environmental conditions. The clarity of the DuraPorter(R) enables the user to visibly check sample integrity, so that it can be opened using the right precautions. Polycarbonate construction and avaiable in 3 colors for user/ ownership identification. Silicone water-tight seal and three lid clasps provide a secure closure. Carry handle and folds neatly into body space allowing the units to be stacked. Lid opens a full 180 ° allowing for total access to contents and making it easier to clean. Will accomodate 13 and 16mm tubes in a 72-place unwire rack or similar sized rack.
Unit is autoclavable.
Unit is autoclavable.
Legal Information
Duraporter is a registered trademark of Heathrow Scientific
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service