Skip to Content
Merck
All Photos(6)

Key Documents

HPA040442

Sigma-Aldrich

Anti-CST7 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-cystatin F (leukocystatin)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

VKGLKYMLEVEIGRTTCKKNQHLRLDDCDFQTNHTLKQTLSCYSEVWVVPWLQHFEVPVLRCH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CST7(8530)

General description

Cystatin-F (CST7) is a glycosylated protein associated with the hematopoietic cells including monocytes and dendritic cells. The CST7 gene is mapped to human chromosome 20p11.21. It belongs to the type II cystatin class.

Immunogen

cystatin F (leukocystatin) recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-CST7 antibody produced in rabbit has been used in immunoblotting and immunohistochemistry.

Biochem/physiol Actions

Cystatin-F (CST7) exists as a dimer. However, its monomeric version after proteolytic cleavage is a cathepsin inhibitor. It effectively inhibits cathepsins C, H, L, K, V and F types. Cystatin F monomeric conversion occurs in the endosomal/lysosomal system. High levels of CST7 expression are observed in the pancreatic ductal adenocarcinoma (PDAC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST81814

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeff D Colbert et al.
Traffic (Copenhagen, Denmark), 10(4), 425-437 (2009-02-05)
Cystatin F is a cysteine protease inhibitor that is selectively expressed in immune cells and unlike other cystatin family members is targeted to a significant extent to intracellular compartments. Initially made as an inactive glycosylated disulfide-linked dimer, cystatin F is
Alexander W Schüttelkopf et al.
The Journal of biological chemistry, 281(24), 16570-16575 (2006-04-08)
Cystatins are important natural cysteine protease inhibitors targeting primarily papain-like cysteine proteases, including cathepsins and parasitic proteases like cruzipain, but also mammalian asparaginyl endopeptidase. Mammalian cystatin F, which is expressed almost exclusively in hematopoietic cells and accumulates in lysosome-like organelles
Mario Nuvolone et al.
PloS one, 12(2), e0171923-e0171923 (2017-02-09)
Misfolding of the cellular prion protein (PrPC) into the scrapie prion protein (PrPSc) results in progressive, fatal, transmissible neurodegenerative conditions termed prion diseases. Experimental and epidemiological evidence point toward a protracted, clinically silent phase in prion diseases, yet there is
PROTEINASE INHIBITORS| Cystatins
Pemberton, PA
A Nurse's Survival Guide to Acute Medical Emergencies null
Milica Perišić Nanut et al.
Frontiers in immunology, 8, 1459-1459 (2017-11-29)
Cystatin F is a cysteine peptidase inhibitor which, unlike other cystatin family members, is targeted to endosomal/lysosomal compartments. It is synthesized as an inactive disulfide-linked dimer which is then converted to an active monomer by proteolytic cleavage of 15 N-terminal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service