Skip to Content
Merck
All Photos(4)

Key Documents

HPA021213

Sigma-Aldrich

Anti-SMARCC2 antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BRG1-associated factor 170, Anti-SWI/SNF complex 170 kDa subunit, Anti-SWI/SNF complex subunit SMARCC2, Anti-SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily C member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

AFGLESSGIAGTTSDEPERIEESGNDEARVEGQATDEKKEPKEPREGGGAIEEEAKEKTSEAPKKDEEKGKEGDSEKESEKSDGDPIVDPEKEKEPKEGQEEVLKEVVESEGERKTKVERDIGEGNLSTAA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMARCC2(6601)

General description

The gene SMARCC2 (SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily C member 2) is mapped to human chromosome 12q13.2. SMARCC2 is commonly referred to as BAF170 (BRG1-associated factor 170).

Immunogen

SWI/SNF complex subunit SMARCC2 recombinant protein epitope signature tag (PrEST)

Application

Anti-SMARCC2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

SWI/SNF (SWItch/Sucrose Non-Fermentable) chromatin-remodeling complexes are important for binding of transcription factors to nucleosomal DNA. SMARCC2 (SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily C member 2) is a core subunit of SWI/SNF. It is suggested that SMARCC2 is required for interaction of SWI/SNF complex with zinc finger DNA-binding domain structures, thereby bringing the complex to nucleosomal sites. SMARCC2 and BAF155 (BRG1-associated factor 155) regulate the protein levels of BAF57 and also support the stoichiometry of the SWI/SNF complex. SMARCC2 is up-regulated in HIV (human immunodeficiency virus)-1 infected cells. Absence of SMARCC2 inhibits viral activation in promyelocytic cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73718

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Stryker et al.
Cancer, 66(7), 1488-1492 (1990-10-01)
A 42-year-old woman developed lower extremity weakness and sensory loss 1 year after external and intracavitary radiotherapy for Stage IB carcinoma of cervix. She has been followed for 5 years posttreatment, and the neurologic abnormalities have persisted, but no evidence
S Kadam et al.
Genes & development, 14(19), 2441-2451 (2000-10-06)
The SWI/SNF family of chromatin-remodeling complexes plays a key role in facilitating the binding of specific transcription factors to nucleosomal DNA in diverse organisms from yeast to man. Yet the process by which SWI/SNF and other chromatin-remodeling complexes activate specific
Jianguang Chen et al.
Molecular and cellular biology, 25(20), 9016-9027 (2005-10-04)
The mammalian SWI/SNF chromatin remodeling complex, whose function is of critical importance in transcriptional regulation, contains approximately 10 protein components. The expression levels of the core SWI/SNF subunits, including BRG1/Brm, BAF155, BAF170, BAF60, hSNF/Ini1, and BAF57, are stoichiometric, with few
Tran Cong Tuoc et al.
Developmental cell, 25(3), 256-269 (2013-05-07)
Increased cortical size is essential to the enhanced intellectual capacity of primates during mammalian evolution. The mechanisms that control cortical size are largely unknown. Here, we show that mammalian BAF170, a subunit of the chromatin remodeling complex mSWI/SNF, is an
Godwin Sokpor et al.
Frontiers in molecular neuroscience, 14, 687581-687581 (2021-07-06)
Radial neuronal migration is a key neurodevelopmental event indispensable for proper cortical laminar organization. Cortical neurons mainly use glial fiber guides, cell adhesion dynamics, and cytoskeletal remodeling, among other discrete processes, to radially trek from their birthplace to final layer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service