Skip to Content
Merck
All Photos(9)

Key Documents

HPA019105

Sigma-Aldrich

Anti-HAPLN1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Cartilage link protein, Anti-Hyaluronan and proteoglycan link protein 1, Anti-LP, Anti-Proteoglycan link protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

DHLSDNYTLDHDRAIHIQAENGPHLLVEAEQAKVFSHRGGNVTLPCKFYRDPTAFGSGIH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HAPLN1(1404)

General description

The gene HAPLN1 (hyaluronan and proteoglycan link protein 1) is mapped to human chromosome 5q14.3. It belongs to HAPLN gene family.

Immunogen

Hyaluronan and proteoglycan link protein 1 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

HAPLN1 (hyaluronan and proteoglycan link protein 1) is a crucial component of the cartilage extracellular matrix. It is responsible for connecting proteoglycans to hyaluronic acid, and thereby regulates compression resistance and shock absorption in the joints. HAPLN1 is down-regulated in Kashin-Beck disease and primary osteoarthritis. Single nucleotide polymorphism in HAPLN1 is linked with spinal osteophyte formation and disc degeneration.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74702

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingjing Zheng et al.
International orthopaedics, 37(10), 2051-2059 (2013-06-12)
Kashin-Beck disease (KBD) is an endemic degenerative osteoarthritis associated with extracellular matrix degradation. The aim of this investigation was to evaluate the role of targeting genes in the pathogenesis of KBD and primary osteoarthritis (OA) involved in extracellular matrix degradation.
Zhiming Lin et al.
Nature genetics, 44(1), 73-77 (2011-12-06)
To identify susceptibility loci for ankylosing spondylitis, we performed a two-stage genome-wide association study in Han Chinese. In the discovery stage, we analyzed 1,356,350 autosomal SNPs in 1,837 individuals with ankylosing spondylitis and 4,231 controls; in the validation stage, we
Tomohiko Urano et al.
European spine journal : official publication of the European Spine Society, the European Spinal Deformity Society, and the European Section of the Cervical Spine Research Society, 20(4), 572-577 (2010-10-19)
Spinal osteoarthritis including disc degeneration is a very common condition in the axial skeletons of aged people. Recently, spinal osteoarthritis has been shown to be influenced by specific genetic risk factors. Vertebral osteophytes, endplate sclerosis, and intervertebral disc narrowing are
Alexandra Naba et al.
Molecular & cellular proteomics : MCP, 11(4), M111-M111 (2011-12-14)
The extracellular matrix (ECM) is a complex meshwork of cross-linked proteins providing both biophysical and biochemical cues that are important regulators of cell proliferation, survival, differentiation, and migration. We present here a proteomic strategy developed to characterize the in vivo
Andrew P Spicer et al.
The Journal of biological chemistry, 278(23), 21083-21091 (2003-03-29)
We describe a vertebrate hyaluronan and proteoglycan binding link protein gene family (HAPLN), consisting of four members including cartilage link protein. The encoded proteins share 45-52% overall amino acid identity. In contrast to the average sequence identity between family members

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service