Skip to Content
Merck
All Photos(3)

Key Documents

HPA018999

Sigma-Aldrich

Anti-GLTSCR2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Glioma tumor suppressor candidate region gene 2 protein, Anti-p60

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
IHC
clone:
polyclonal
species reactivity:
human
citations:
12
technique(s):
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

REAEADKPRRLGRLKYQAPDIDVQLSSELTDSLRTLKPEGNILRDRFKSFQRRNMIEPRERAKFKRKYKVKLVEKRAFREIQL

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GLTSCR2(29997)

General description

The gene GLTSCR2 (glioma tumor suppressor candidate region gene 2 protein) is mapped to human chromosome 19q. The protein localizes in the nucleolus and the nucleoplasm. GLTSCR2 is also called as PICT1 (PTEN carboxyl terminus 1).

Immunogen

Glioma tumor suppressor candidate region gene 2 protein recombinant protein epitope signature tag (PrEST)

Application

Anti-GLTSCR2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

GLTSCR2 (glioma tumor suppressor candidate region gene 2) regulates expression of tumor suppressor p53. It suppresses transfer of ribosomal protein L11 to the nucleoplasm. In the nucleoplasm, L11 inhibits E3 ubiquitin-protein ligase, MDM2 (Mouse double minute 2), resulting in p53 activation. In presence of ribosomal stress GLTSCR2 moves to the nucleoplasm and stabilizes p53, thereby inhibiting cell cycle progression. GLTSCR2 plays an important role in cellular respiration. GLTSCR2 is down-regulated in cervical cancer, squamous cell carcinoma, prostatic adenocarcinoma and breast cancer. On the other hand, GLTSCR2 is involved in gastric cancer progression.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74821

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomohiko Maehama et al.
The Journal of biological chemistry, 289(30), 20802-20812 (2014-06-14)
The nucleolar protein PICT1 regulates tumor suppressor p53 by tethering ribosomal protein L11 within the nucleolus to repress the binding of L11 to the E3 ligase MDM2. PICT1 depletion results in the release of L11 to the nucleoplasm to inhibit
Jee-Youn Kim et al.
Cancer letters, 340(1), 134-140 (2013-08-08)
GLTSCR2 is a nuclear/nucleolar protein that translocates to the nucleoplasm, suppressed and mutated in human cancers. Our aim in this study was to investigate whether downregulation or cytoplasmic expression of GLTSCR2 has any pathological significance in prostatic cancer development or
Kyoko Okamura et al.
Oncoscience, 1(5), 375-382 (2015-01-17)
PICT1 is a key regulator of the MDM2-TP53 pathway. High mRNA expression levels of PICT1 are associated with poor prognosis in several cancers with wild-type TP53. In this study, we identified the PICT1 protein expression profile in non-small cell lung
Ahrim Moon et al.
Pathology, research and practice, 209(11), 700-704 (2013-09-24)
Glioma tumor-suppressor candidate region gene2 (GLTSCR2) is a recently identified nucleolus-localized protein participating in the regulation of cell cycle progression and apoptosis. Down-regulation of GLTSCR2 in several types of cancers and increased transforming activity in GLTSCR2-downregulated cancer cells indicated its
Jiawen Zhang et al.
Archives of gynecology and obstetrics, 291(2), 413-418 (2014-08-15)
GLTSCR2 was originally identified as a candidate tumor suppressor in several types of cancers. The present study was to investigate the expression pattern of GLTSCR2 in different cervical lesion tissues, appraise its potential role in cervical cancerogenesis. 225 histologically confirmed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service