Skip to Content
Merck
All Photos(3)

Key Documents

HPA014864

Sigma-Aldrich

Anti-KCNS3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Delayed-rectifier K(+) channel alpha subunit 3, Anti-Potassium voltage-gated channel subfamily S member 3, Anti-Voltage-gated potassium channel subunit Kv93

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

KDIDVDQCSEDAPEKCHELPYFNIRDIYAQRMHAFITSLSSVGIVVSDPDSTDASSIEDNEDICNTTSLENCTA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNS3(3790)

General description

KCNS3 (potassium voltage-gated channel, modifier subfamily S, member 3) codes for the modulatory α-subunit of Kv9.3 votlage-gated potassium channel. This gene is found to be expressed in vascular tissues of human placenta and syncytiotrophoblast. It is localized to human chromosome 2, and has a wide range of tissue expression, with the highest being in brain and lungs.

Immunogen

Potassium voltage-gated channel subfamily S member 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

KCNS3 (potassium voltage-gated channel, modifier subfamily S, member) controls the tonicity and sensitivity of neural and muscle cells, by regulating their resting potential. It controls the membrane potential at sub-threshold levels. It is involved in the repetitive firing and high-frequency of parvalbumin neurons, by controlling the repolarization of action potential. In parvalbumin neurons, it is responsible for the exact recognition of co-incident excitatory synaptic inputs, which in turn affects γ-oscillations. In schizophrenia, KCNS3 expression is lowered in prefrontal cortical parvalbumin neurons, which might lead to defective cognitive processes. Variants in this gene are associated with airway hyperresponsiveness. It forms hetero-tetrameric channel with Kv2.1 α-subunits, and this channel contributes to the myogenic regulation of arterial diameter of the cerebrum.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73064

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L C Fu et al.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas, 50(11), e6237-e6237 (2017-09-14)
Intrauterine growth retardation (IUGR) is associated with the development of adult-onset diseases, including pulmonary hypertension. However, the underlying mechanism of the early nutritional insult that results in pulmonary vascular dysfunction later in life is not fully understood. Here, we investigated
Xi Zoë Zhong et al.
The Journal of physiology, 588(Pt 22), 4519-4537 (2010-09-30)
Cerebral vascular smooth muscle contractility plays a crucial role in controlling arterial diameter and, thereby, blood flow regulation in the brain. A number of K(+) channels have been suggested to contribute to the regulation of diameter by controlling smooth muscle
Ke Hao et al.
Human genetics, 116(5), 378-383 (2005-02-17)
Airway hyperresponsiveness (AHR) is one of the major clinical symptoms and intermediate phenotypes of asthma. A recent genome-wide search for asthma quantitative trait loci has revealed a significant linkage signal between a p-terminal region of chromosome 2 and AHR. Thus
G K Fyfe et al.
Journal of obstetrics and gynaecology : the journal of the Institute of Obstetrics and Gynaecology, 32(7), 624-629 (2012-09-05)
Human placental expression of K(V)9.3, a voltage-gated K channel linked to tissue oxygenation responses, has been suggested at the messenger RNA level but tissue localisation has not been described. We aimed to: (1) produce an antibody to human K(V)9.3 and
Ying Lv et al.
American journal of physiology. Lung cellular and molecular physiology, 305(11), L856-L865 (2013-10-01)
Chronic hypoxia pulmonary hypertension (CH-PHT) in adulthood is likely to be of fetal origin following intrauterine growth retardation (IUGR). Oxygen (O₂)-sensitive voltage-gated potassium channels (Kv channels) in resistance pulmonary artery smooth muscle cells (PASMCs) play an important role in scaling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service