Skip to Content
Merck
All Photos(2)

Key Documents

HPA010549

Sigma-Aldrich

Anti-WTAP antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Female-lethal(2)D homolog, Anti-Pre-mRNA-splicing regulator WTAP, Anti-WT1-associated protein, Anti-Wilms tumor 1-associating protein, Anti-hFL(2)D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect immunofluorescence: 0.25-2 μg/mL
western blot: 0.04-0.4 μg/mL

immunogen sequence

TNEEPLPKKVRLSETDFKVMARDELILRWKQYEAYVQALEGKYTDLNSNDVTGLRESEEKLKQQQQESARRENILVMRLATKEQEMQECTTQIQYLKQVQQPSVA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WTAP(9589)

General description

WTAP (Wilms tumor 1 associated protein) is a highly conserved protein that functions as an interaction partner of WT1 protein, and acts as a switch. It is exclusively localized to the nucleus, and was first recognized in a kidney cell line, as a WT1-associated protein. It has a molecular weight of 20kDa.

Immunogen

Pre-mRNA-splicing regulator WTAP recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

WTAP (Wilms tumor 1 associated protein), along with WT1, controls the balance between quiescence and proliferation. It functions as an oncogene in gliomas and acute myeloid leukemia. It negatively controls the proliferation of vascular smooth muscle cells (SMCs), where its expression is elevated as SMCs achieve replicative quiescence. This protein is up-regulated in cholangiocarcinoma, where it is responsible for the control of migration and invasiveness. This protein controls the activity of EGFR (epidermal growth factor), and is up-regulated in glioblastomas, where it controls the migration and invasion.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71499

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

WTAP is a novel oncogenic protein in acute myeloid leukemia.
H Bansal et al.
Leukemia, 28(5), 1171-1174 (2014-01-15)
Du-Il Jin et al.
Cancer science, 103(12), 2102-2109 (2012-09-11)
Glioblastoma is a diffusely growing malignant brain tumor and among the most aggressive of all tumors. Wilms' tumor 1-associating protein (WTAP) is a nuclear protein that has been associated with regulation of proliferation and apoptosis. Although its dynamic expression and
Theodore W Small et al.
Circulation research, 99(12), 1338-1346 (2006-11-11)
Smooth muscle cells (SMCs) are called on to proliferate during vascular restructuring but must return to a nonproliferative state if remodeling is to appropriately terminate. To identify mediators of the reacquisition of replicative quiescence, we undertook gene expression screening in
Hong-Jae Jo et al.
Journal of gastroenterology, 48(11), 1271-1282 (2013-01-29)
Wilms' tumor 1-associating protein (WTAP) is a nuclear protein that has been associated with the regulation of proliferation and apoptosis. Although its dynamic expression and physiological functions in vascular cells have been reported, its expression and roles in cholangiocarcinoma cells
Gaocai Li et al.
Cell death & disease, 11(2), 103-103 (2020-02-08)
N6 methyladenosine (m6A) is one of the most prevalent epitranscriptomic modifications of mRNAs, and plays a critical role in various bioprocesses. Bone-derived mesenchymal stem cells (BMSCs) can attenuate apoptosis of nucleus pulposus cells (NPCs) under compression; however, the underlying mechanisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service