Skip to Content
Merck
All Photos(8)

Key Documents

HPA002868

Sigma-Aldrich

Anti-SDHB antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-SDH, Anti-SDH1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

EGKQQYLQSIEEREKLDGLYECILCACCSTSCPSYWWNGDKYLGPAVLMQAYRWMIDSRDDFTEERLAKLQDPFSLYRCHTIMNCTRTCPKGLNPGKAIAEIKKMMATY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SDHB(6390)

Looking for similar products? Visit Product Comparison Guide

General description

Complex II (succinate-ubiquinone oxidoreductase) is a mitochondrial enzyme complex that regulates aerobic respiration and TCA cycle. It is the smallest mitochondrial respiratory chain complex and consists of four subunits, namely, SDHA, SDHB, SDHC, and SDHD. Germline mutations in SDHB have been associated with invasive paragangliomas and pheochromocytomas . Anti-SDHB antibody is specific for SDHB in humans.
Succinate dehydrogenase complex iron sulfur subunit B is a catalytic core subunit of a heterodimeric mitochondrial enzyme, succinate dehydrogenase (SDH). The hydrophilic SDHB subunit is exposed to the matrix side of the inner mitochondrial membrane. The SDHB gene, spanning 35.4 Kb, with eight exons, is located on human chromosome 1p36.13. The gene encodes for a 280 amino-acid protein.

Immunogen

Succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-SDHB antibody produced in rabbit has been used in western blotting and immunohistochemistry.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Western Blotting (1 paper)

Biochem/physiol Actions

Succinate dehydrogenase complex iron sulfur subunit B plays a key role in cellular metabolism and ATP synthesis by linking the biochemical reactions involved in oxidation of glucose to the mitochondrial electron transport chain. It is also responsible for the conversion of succinate to fumarate as part of the Kreb′s cycle. Germline mutations in the gene is associated with pheochromocytoma/paraganglioma syndrome, an autosomal dominant disorder.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86628

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B E Baysal et al.
Journal of molecular medicine (Berlin, Germany), 79(9), 495-503 (2001-11-03)
This review presents our current knowledge on the genetic and phenotypic aspects of mitochondrial complex II gene defects. The mutations of the complex II subunits cause two strikingly different group of disorders, revealing a phenotypic dichotomy. Genetic disorders of the
Succinate dehydrogenase B (SDHB) immunohistochemistry for the evaluation of muscle biopsies
Punsoni M, et al.
Applied Immunohistochemistry & Molecular Morphology, 25(9), 645-650 (2017)
A novel succinate dehydrogenase type B mutation in an Iranian family. Its genetic and clinical evaluation
Ghazi AA, et al.
Hormones (Athens, Greece), 13(4), 568-573 (2014)
Cécile Nozières et al.
European journal of endocrinology, 166(6), 1107-1111 (2012-03-21)
Ten percent of paragangliomas are malignant and one-third occurs in a genetic background. We report a case of succinate dehydrogenase subunit B (SDHB)-related malignant paraganglioma with dramatic response to temozolomide and capecitabine regimen (decrease in tumor size of 70% with
Functional consequences of a SDHB gene mutation in an apparently sporadic pheochromocytoma
Gimenez-Roqueplo AP, et al.
The Journal of Clinical Endocrinology and Metabolism, 87(10), 4771-4774 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service