Skip to Content
Merck
All Photos(1)

Key Documents

H0665

Sigma-Aldrich

Monoclonal Anti-Hepatocyte Growth Factor antibody produced in mouse

clone 24516, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-HGF

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

24516, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

capture ELISA: 2-8 μg/mL
western blot: 1-2 μg/mL

isotype

IgG1

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL4(3565)

Looking for similar products? Visit Product Comparison Guide

General description

Hepatocyte Growth Factor (HGF) is a multifunctional, heparin-binding factor that stimulates the proliferation of liver cells. The cellular effects of HGF are mediated by its receptor, c-Met, that leads to significant activation of ERK, Src/FAK, STAT3 and PI3K signaling. Cells of mesenchymal origin express HGF that stimulates the c-Met expression via an autocrine loop. Various functions of the HGF/c-Met signaling include proliferation, cell survival, motility and adhesion and tissue regeneration. Apart from liver, HGF also mediates regeneration of kidney, lung and gastro-intestines. It is also important for maintaining the insulin production and prevents hypoglycemia, self-repair of cartilage, muscle and skin and hematopoiesis. The HGF/c-Met signaling is reported as a promising therapeutic target in prevention of angiogenesis in cancer and maintaining synaptic plasticity in Alzheimer′s disease
Monoclonal Anti-Hepatocyte Growth Factor recognizes human HGF. The antibody shows less that 0.25% cross-reactivity with recombinant human Macrophage Stimulating Protein (MSP).

Immunogen

purified, Sf21-derived recombinant human hepatocyte growth factor.

Application

Anti-Hepatocyte Growth Factor antibody may be used for ELISA at a concentration range of 2-8 μg/ml. For immunoblotting, a working concentration of 1-2 μg/ml is recommended.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunsun Dai et al.
The Journal of biological chemistry, 278(29), 27080-27087 (2003-05-15)
Type I diabetes is an autoimmune disease that results in destructive depletion of the insulin-producing beta cells in the islets of Langerhans in pancreas. With the knowledge that hepatocyte growth factor (HGF) is a potent survival factor for a wide
Shiv Sharma
TheScientificWorldJournal, 10, 457-461 (2010-03-23)
The hepatocyte growth factor (HGF) was initially identified as a protein that promoted growth of hepatocytes. It regulates proliferation and survival of different types of cells. HGF signaling, which is initiated by its binding to a receptor tyrosine kinase, plays
H Ohmichi et al.
The American journal of physiology, 270(6 Pt 1), L1031-L1039 (1996-06-01)
Hepatocyte growth factor (HGF) has mitogenic, morphogenic, and motogenic activities on epithelial cells and plays important roles in regeneration of the liver and the kidney. We previously found that the expression of HGF gene is rapidly induced in the lung
Toshikazu Nakamura et al.
Proceedings of the Japan Academy. Series B, Physical and biological sciences, 86(6), 588-610 (2010-06-17)
It has been more than 25 years since HGF was discovered as a mitogen of hepatocytes. HGF is produced by stromal cells, and stimulates epithelial cell proliferation, motility, morphogenesis and angiogenesis in various organs via tyrosine phosphorylation of its receptor
K Matsumoto et al.
Critical reviews in oncogenesis, 3(1-2), 27-54 (1992-01-01)
Hepatocyte growth factor (HGF) is the most potent mitogen for mature hepatocytes and seems to act as a hepatotropic factor that has not been purified over the past 30 years. HGF was first purified from rat platelets in 1986. HGF

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service