Skip to Content
Merck
All Photos(2)

Key Documents

H0627

Sigma-Aldrich

6-Hydroxymelatonin

Synonym(s):

3-(N-Acetylaminoethyl)-6-hydroxy-5-methoxyindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N2O3
CAS Number:
Molecular Weight:
248.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

solubility

alcohol: soluble

Quality Level

SMILES string

COc1cc2c(CCNC(C)=O)c[nH]c2cc1O

InChI

1S/C13H16N2O3/c1-8(16)14-4-3-9-7-15-11-6-12(17)13(18-2)5-10(9)11/h5-7,15,17H,3-4H2,1-2H3,(H,14,16)

InChI key

OMYMRCXOJJZYKE-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

6-Hydroxymelatonin (6-OHM) is a melatonin metabolite. It is produced in the liver by the action of cytochrome P450 enzyme as well as by photodegradation of melatonin. In the central nervous system, it exists as a sulfated form. 6-OHM is a partial melatonin receptor MT2 agonist.

Application

6-Hydroxymelatonin has been used as a melatonin derivative to test its protective effects in ultra violet B (UVB)-induced oxidative stress melanocytes and keratinocytes.

Biochem/physiol Actions

6-Hydroxymelatonin (6-OHM) is an antioxidant with a free radical scavenging role. Like melatonin, it reduces the impact of UVB-induced oxidative stress in melanocytes. It also aids protection during iron (Fe2+)-induced neurotoxicity. 6-OHM effectively reduces lipid peroxidation and superoxide anion production induced by potassium cyanide (KCN).

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Herwig Frisch et al.
The Journal of clinical endocrinology and metabolism, 89(9), 4388-4390 (2004-09-10)
Melatonin (MLT), the pineal gland hormone involved in the regulation of circadian rhythms, shows characteristic diurnal variation. Its physiological role in humans is not clear. Exposure to high altitudes may disrupt the circadian rhythm and lead to various endocrine changes.
Zorica Janjetovic et al.
Journal of pineal research, 57(1), 90-102 (2014-05-29)
We investigated the protective effects of melatonin and its metabolites: 6-hydroxymelatonin (6-OHM), N1-acetyl-N2-formyl-5-methoxykynuramine (AFMK), N-acetylserotonin (NAS), and 5-methoxytryptamine (5-MT) in human keratinocytes against a range of doses (25, 50, and 75 mJ/cm2) of ultraviolet B (UVB) radiation. There was significant
S Härtter et al.
Clinical pharmacology and therapeutics, 70(1), 10-16 (2001-07-14)
Melatonin is a hormone that is metabolized by cytochrome P450 (CYP) 1A2 to its main primary metabolite 6-hydroxymelatonin. We therefore evaluated the utility of oral melatonin as a marker of hepatic CYP1A2 activity. Twenty-five milligrams of melatonin was given at
Katsuhisa Sakano et al.
Biochemical pharmacology, 68(9), 1869-1878 (2004-09-29)
Melatonin, an indolic pineal hormone, is produced primarily at night in mammals and is important in controlling biological rhythms. Although melatonin is known to be effective as a free radical scavenger and has an anti-cancer effect, carcinogenic properties have also
K N Ting et al.
British journal of pharmacology, 122(7), 1299-1306 (1998-01-08)
1. In this study we compared the vasoconstrictor activity of melatonin in rat isolated tail artery using two different recording systems, the Halpern pressure myograph and the Halpern-Mulvany wire myograph, with the view to determining a reliable method for obtaining

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service