Skip to Content
Merck
All Photos(1)

Documents

GW20090F

Sigma-Aldrich

Anti-VLDL antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-20090F

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VLDLR(7436)

Related Categories

Application

Anti-VLDL antibody produced in chicken is suitable for indirect ELISA and western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

The very-low-density-lipoprotein receptor (VLDLR) is a transmembrane lipoprotein receptor and belongs to low-density-lipoprotein (LDL) receptor family. It is encoded by the VLDLR gene in humans. The LDL receptor gene family members are involved in ligand-mediated signal transduction. VLDLR is involved in the Reelin signaling pathway helping in neuronal migration in the cerebral cortex and cerebellum. It is strongly induced during adipogenesis and actively participates in adipocyte hypertrophy. VLDLR contributes to adipose tissue inflammation and mediates VLDL-induced lipid accumulation. It induces inflammation and ER stress in adipocytes and macrophages. VLDL receptor also plays a role in signaling processes.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew Nguyen et al.
The Journal of biological chemistry, 289(3), 1688-1703 (2013-12-03)
Obesity is associated with adipose tissue remodeling, characterized by adipocyte hypertrophy and macrophage infiltration. Previously, we have shown that very low density lipoprotein receptor (VLDLR) is virtually absent in preadipocytes but is strongly induced during adipogenesis and actively participates in
J Nimpf et al.
Biochimica et biophysica acta, 1529(1-3), 287-298 (2000-12-09)
The discovery of an ever growing number of low density lipoprotein (LDL) receptor gene family members has triggered research into many different directions. Here we first summarize the results of classical studies on the role of the LDL receptor in
Praseetha Kizhakkedath et al.
Biochimica et biophysica acta, 1843(12), 2871-2877 (2014-09-01)
Dysequilibrium syndrome (DES, OMIM 224050) is a genetically heterogeneous condition that combines autosomal recessive non-progressive cerebellar ataxia with mental retardation. The subclass dysequilibrium syndrome type 1 (CAMRQ1) has been attributed to mutations in the VLDLR gene encoding the very low

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service