Skip to Content
Merck
All Photos(1)

Documents

GUTMLS01

Sigma-Aldrich

Microbiome Metabolite Library Of Standards Plate 1 (Ethanol And Methanol Soluble)

Supplied by IROA Technologies

Synonym(s):

GUTMLS, Mass Spectrometry Metabolite Library

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

Quality Level

packaging

pkg of 1 plates

manufacturer/tradename

IROA Technologies

plate size

96 wells

shipped in

dry ice

storage temp.

−20°C

General description

GUTMLS Plate 1 is a collection of high quality small biochemical molecules that span a broad range of primary metabolism. These are high purity (>95%) compounds supplied in an economical, ready-to-use format. This library of standards is most commonly used for metabolite qualification and quantification for MS and NMR, functional cellular assays and phenotypic screening. Metabolite descriptors provided with product include: Name, Parent CID, molecular formula, molecular weight, CAS, ChEBI, HMDB ID, PubChem Compound and Substance ID. The library is intended to be used for mass spectrometry metabolomics applications and provides a broad representation of primary metabolism.

Application

Metabolite qualification and quantification for MS, NMR, etc.; Functional cellular assays; Phenotypic screening

Features and Benefits

To view a complete list of compounds in the library visit GUTMLS Plate Map. The complete plate map with name, Parent CID, molecular formula, molecular weight, CAS, ChEBI, HMBD ID, PubChem Compound and Substance ID are provided with the software dowmload. Broad metabolite spectrum, key primary metabolites and intermediates covering key metabolic pathways, including the following classes of compounds: Carboxylic acids, amino acids; Biogenic amines, polyamines; Nucleotides, coenzymes and vitamins; Mono-and disaccharides; Fatty acids, lipids, steroids and hormones

Suitability

Suitable for manual and automated work flows

Preparation Note

GUTMLS Plate 1 compounds are conveniently provided at 5μg per well, enough for multiple injections. Compounds can either be used as standards and injected individually or a strategy may be employed to pool compounds by row to allow multiple mixtures to be analyzed per injection. For more information on GUTMLS01, solubilization and preparation notes please refer to the product information sheet at the top of the page.

Other Notes

For additional information on our range of Biochemicals, please complete this form.
MLSDiscovery software package is distributed with and is tailored to work with GUTMLS01. A user manual and video instructions are provided.

To download the software package, click here - MLSDiscovery software.

To view the MLSDiscovery Video visit: MLSDiscovery Tutorial Video.

Legal Information

GUTMLS is a trademark of IROA Technologies LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

We provide small molecule metabolite libraries and kits with a comprehensive range of primary metabolite compounds covering key metabolic pathways for your metabolomics research.

We provide small molecule metabolite libraries and kits with a comprehensive range of primary metabolite compounds covering key metabolic pathways for your metabolomics research.

We provide small molecule metabolite libraries and kits with a comprehensive range of primary metabolite compounds covering key metabolic pathways for your metabolomics research.

We provide small molecule metabolite libraries and kits with a comprehensive range of primary metabolite compounds covering key metabolic pathways for your metabolomics research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service