Recommended Products
form
solution
shelf life
Expiry date on the label.
IVD
for in vitro diagnostic use
concentration
6 g/L
application(s)
hematology
histology
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
Application
Gill formulation No. 3 may be used for cytology and/or histology as a progressive or regressive stain depending on length of staining time.. Used with hematoxylin and eosin staining.
Other Notes
6 g/L certified hematoxylin
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - STOT RE 2 Oral
Target Organs
Kidney
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Stem cell research, 37, 101444-101444 (2019-05-11)
Human pluripotent stem cells (hPSCs) acquire changes at the genomic level upon proliferation and differentiation (Peterson and Loring, 2014). Studies from International Stem Cell Initiative and independent laboratories identified a copy number variant (CNV) in hES cell lines displaying a
The Prostate, 79(9), 980-993 (2019-04-19)
Prostatic inflammation and various proinflammatory systemic comorbidities, such as diabetes and obesity are associated with human benign prostatic hyperplasia (BPH). There is a paucity of in vivo models reflecting specific aspects of BPH pathogenesis. Our aim was to investigate the
Photochemistry and photobiology, 96(4), 863-869 (2019-12-04)
Tranexamic acid (TSA) is widely used as an antiaging treatment for reducing melasma and wrinkles. There are various mechanisms for wrinkle formation, and one of them is due to damage of the mitochondria. Research on mitochondria in the skin is
Frontiers in endocrinology, 11, 586980-586980 (2020-11-17)
Sterol 27-hydroxylase (CYP27A1) is a key enzyme in bile acids (BAs) biosynthesis and a regulator of cholesterol metabolism. Cyp27a1/Apolipoprotein E double knockout (DKO) mice fed with western diet (WD) are protected from atherosclerosis via up-regulation of hepatic Cyp7a1 and Cyp3a11.
Molecular cancer therapeutics, 18(2), 421-436 (2018-11-14)
Systematic approaches for accurate repurposing of targeted therapies are needed. We developed and aimed to biologically validate our therapy predicting tool (TPT) for the repurposing of targeted therapies for specific tumor types by testing the role of Bromodomain and Extra-Terminal
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service