Skip to Content
Merck
All Photos(1)

Key Documents

GE29401322

Sigma-Aldrich

StrepTrap XT prepacked chromatography column

pkg of 1 × 5 mL, prepacked chromatography column

Synonym(s):

StrepTrap High Performance

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106501
NACRES:
SB.53
Pricing and availability is not currently available.

grade

for chromatography

packaging

pkg of 1 × 5 mL

manufacturer/tradename

Cytiva 29401322

Looking for similar products? Visit Product Comparison Guide

General description

A chromatography resin for the purification of recombinant proteins tagged with Strep-tag II and Twin-Strep-tag, which provides higher affinity, resulting in higher yields and lower loss of valuable target protein.

Features and Benefits

StrepTrap XT is a ready to use column prepacked with Strep-Tactin XT Sepharose providing the benefits:
  • Highly pure Strep-tag II and Twin-Strep-tag recombinant proteins.
  • Fast purifications with simple elution.
  • Physiological conditions and mild elution preserve target protein activity.
  • Fast and easy regeneration.
  • Compatible with a wide range of detergents, denaturants, and other additives.

Legal Information

StrepTrap is a trademark of Cytiva

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service