Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5318-06

Ni Sepharose 6 Fast Flow

Cytiva 17-5318-06, pack of 5 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56
Pricing and availability is not currently available.

packaging

pack of 5 mL

manufacturer/tradename

Cytiva 17-5318-06

matrix

6% cross-linked agarose

particle size

45-165 μm

average diameter

90 μm

cleaning in place

2-14(Ni2+-stripped medium.)

working range

3-12(Ni2+-stripped medium.)

capacity

~40 mg binding capacity(histidine-tagged protein)

suitability

suitable for bioprocess medium

General description

Ni Sepharose 6 Fast FLow is a BioProcess medium that combines the advantages of Ni?? for purification of histidine tagged proteins, with the well industrially established properties of the Sepharose Fast FLow platform.

Application

Ni Sepharose 6 Fast FLow is a member of the Cytiva range of IMAC chromatography media for capture and intermediate purification.

Ni Sepharose 6 Fast FLow is composed of cross-linked 6% Agarose beads modified with a novel chelating ligand immobilized to the base matrix and pre-charged with Ni2? ions for purification of histidine tagged Proteins.

The coupling chemistry and the novel ligand is designed to give minimal ligand and Ni2? leakage while providing high capacity.

Ni Sepharose 6 Fast FLow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Ni Sepharose 6 Fast FLow meets industrial demands with security of supply and comprehensive technical and regulatory support.

Features and Benefits

  • Designed for scaling up purification of histidine-tagged Proteins
  • High Protein binding capacity and minimal leakage of Ni2? ions
  • As BioProcess medium it fulfills industrial demands for security of supply, robust performance and regulatory support.
  • Available in convenient prepacked HisPrep and HisTrap column formats.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

IMAC Sepharose products, Ni Sepharose products and Fe Sepharose products
These products are sold under a license from Sigma-Aldrich under patent number EP 1276716 (Metal chelating compositions) and equivalent patents and patent applications in other countries.
HisPrep is a trademark of Cytiva
HisTrap is a trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pär Comstedt et al.
PloS one, 9(11), e113294-e113294 (2014-11-20)
There is currently no Lyme borreliosis vaccine available for humans, although it has been shown that the disease can be prevented by immunization with an OspA-based vaccine (LYMErix). Outer surface protein A (OspA) is one of the dominant antigens expressed

Articles

This page shows high-throughput screening using His MultiTrap™ HP and His MultiTrap™ FF 96-well filter plates from Cytiva.

Protocols

HisTrap FF Kit purifies histidine-tagged proteins directly from cell lysates, minimizing sample pretreatment for rapid purification.

HisTrap FF Kit purifies histidine-tagged proteins directly from cell lysates, minimizing sample pretreatment for rapid purification.

HisTrap FF Kit purifies histidine-tagged proteins directly from cell lysates, minimizing sample pretreatment for rapid purification.

HisTrap FF Kit purifies histidine-tagged proteins directly from cell lysates, minimizing sample pretreatment for rapid purification.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service