GE17-1043-01
Superdex® 200 Prep Grade
Cytiva 17-1043-01, pack of 150 mL
Synonym(s):
Superdex 200 Column
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41115709
NACRES:
NA.56
Recommended Products
General description
Superdex® 200 prep grade gel filtration medium offers very high resolution fractionation for separation, polishing and product formulation of monoclonal antibodies.
Superdex® prep grade is a preparative gel filtration medium with a composite matrix of dextran and agarose. This matrix combines the excellent gel filtration properties of cross-linked dextran (Sephadex) with the physical and chemical stabilities of highly cross-linked agarose, to produce a separation medium with outstanding selectivity and high resolution. In addition, its low non-specific interaction permits high recovery of biological material. Together these properties make Superdex® prep grade the first choice gel filtration media for all applications from laboratory to process scale.
Superdex® prep grade is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development.
As a BioProcess medium, Superdex® prep grade meets industrial demands with security of supply and comprehensive technical and regulatory support.
Superdex® prep grade is available in three selectivities:
Superdex® 30 prep grade is designed for purification of molecules in the molecular weight range up to 10 000 MW, such as peptides, oligosaccharides and small proteins.
Superdex® 75 prep grade is recommended for the purification of recombinant proteins. (Fractionation range 3000-70000 Da).
Superdex® 200 prep grade is recommended for the purification of monoclonal antibodies. (Fractionation range 10 000 -600 000 Da)
Superdex® prep grade is a preparative gel filtration medium with a composite matrix of dextran and agarose. This matrix combines the excellent gel filtration properties of cross-linked dextran (Sephadex) with the physical and chemical stabilities of highly cross-linked agarose, to produce a separation medium with outstanding selectivity and high resolution. In addition, its low non-specific interaction permits high recovery of biological material. Together these properties make Superdex® prep grade the first choice gel filtration media for all applications from laboratory to process scale.
Superdex® prep grade is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development.
As a BioProcess medium, Superdex® prep grade meets industrial demands with security of supply and comprehensive technical and regulatory support.
Superdex® prep grade is available in three selectivities:
Superdex® 30 prep grade is designed for purification of molecules in the molecular weight range up to 10 000 MW, such as peptides, oligosaccharides and small proteins.
Superdex® 75 prep grade is recommended for the purification of recombinant proteins. (Fractionation range 3000-70000 Da).
Superdex® 200 prep grade is recommended for the purification of monoclonal antibodies. (Fractionation range 10 000 -600 000 Da)
Features and Benefits
- Gel filtration medium providing exceptional resolution with short run times and good recovery.
- Superdex® 200 prep grade is recommended for the purification of monoclonal antibodies.
- BioProcess medium supported for industrial applications and well-established in approved processes.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Superdex is a registered trademark of Cytiva
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service