GE17-0310-10
Ficoll® PM 70
Cytiva 17-0310-10, pack of 100 g
Synonym(s):
Density Gradient Solution, Ficoll Solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352201
NACRES:
NA.25
Recommended Products
form
solid
spray-dried
packaging
pack of 100 g
manufacturer/tradename
Cytiva 17-0310-10
color
White
solubility
soluble
Related Categories
General description
High molecular weight sucrose-polymers formed by copolymerization of sucrose with epichlorohydrin. The molecules are highly branched and the high content of hydroxyl groups leads to very good solubility in aqueous media.
It is delivered as a spray-dried powder and is therefore readily soluble in aqueous media when it is added slowly with concomitant stirring. Any lumps formed during this procedure will dissolve fairly quickly. Concentrations up to 50% (w/v) can easily be attained
Features and Benefits
- Displays minimal toxicity
- Has favorable viscosity and osmotic properties
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C ()
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Ficoll is a registered trademark of Cytiva
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service