Key Documents
GBL664273
Grace Bio-Labs CultureWell silicone isolator sheet material
thickness 1.6 mm, (No adhesive)
Synonym(s):
silicone sheet
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
Pricing and availability is not currently available.
material
clear silicon
feature
(No adhesive)
packaging
pack of 5 ea
manufacturer/tradename
Grace Bio-Labs 664273
L × W
180 mm × 130 mm
thickness
1.6 mm
General description
CultureWell clear silicone sheet material allows researchers to create their own removable hydrophobic barriers to isolate specimens. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides.
Legal Information
Culturewell is a trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Paula G Miller et al.
Biotechnology and bioengineering, 117(2), 486-497 (2019-10-15)
Efficient and economical delivery of pharmaceuticals to patients is critical for effective therapy. Here we describe a multiorgan (lung, liver, and breast cancer) microphysiological system ("Body-on-a-Chip") designed to mimic both inhalation therapy and/or intravenous therapy using curcumin as a model