Key Documents
GBL622304
Grace Bio-Labs CoverWell perfusion chambers
4 chambers, port diam. 1.5 mm, chamber W × L × thickness 6.35 mm × 19 mm × 2.5 mm
Synonym(s):
leak proof enclosure for slides, microscope stick on multiwell chamber, reusable press to seal multiwell chamber, water tight multiple well chamber
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
Pricing and availability is not currently available.
material
clear polycarbonate backing
red silicon wells
sterility
non-sterile
feature
wells 4
packaging
pack of 20 ea
manufacturer/tradename
Grace Bio-Labs 622304
chamber W × L × thickness
6.35 mm × 19 mm × 2.5 mm
port diam.
1.5 mm
wells
4
General description
CoverWell perfusion chambers press-to-seal covers form water-tight, multiwell cell incubation or cytochemistry chambers when pressed to coverslips or microscope slides. Reagents can be quickly added and removed through dual access ports without disturbing or cross-contaminating specimens. Chambers are available in five chamber configurations allowing up to eight different specimens or analytical tests on one coverglass.
CoverWell perfusion chambers form removable, water-tight multiwell enclosures by pressing to coverglass or microscope slides. Chambers are made of medical-grade silicone bonded to clear, UV-transparent plastic, and are removable for permanent mounting. The thin polycarbonate cover displays excellent transparency. The perfusion chambers are ideal for live cell imaging on upright or inverted microscopes at high magnification.
Legal Information
CoverWell is a trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)